Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 115870058-115870283 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP3S1
Entrez Gene ID
1176
Gene Name
adaptor related protein complex 3 sigma 1 subunit
Gene Aliases
CLAPS3, Sigma3A
UniGene
Hs.406191
Gene Chromosome Location
Chr.5: 115841606-115914081 on Build GRCh38
Transcripts
NM_001002924.2 NM_001284.3 NM_001318090.1 NM_001318091.1 NM_001318093.1 NM_001318094.1 XM_011543136.2 XM_017009024.1 XM_017009025.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1059872 c.209C>T p.T70M Chr.5: 115870064-115870064 on Build GRCh38
1671817 c.236C>A p.S79Y Chr.5: 115870091-115870091 on Build GRCh38
6101374 c.262G>T p.D88Y Chr.5: 115870117-115870117 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs750434158 Chr.5: 115870131-115870132 on Build GRCh38
rs747540923 Chr.5: 115870133-115870134 on Build GRCh38
rs1043666010 Chr.5: 115870249-115870249 on Build GRCh38
rs1039561565 Chr.5: 115870193-115870193 on Build GRCh38
rs917249600 Chr.5: 115870126-115870126 on Build GRCh38
rs768946863 Chr.5: 115870142-115870142 on Build GRCh38
rs781293527 Chr.5: 115870110-115870110 on Build GRCh38
rs773895194 Chr.5: 115870145-115870145 on Build GRCh38
rs779059797 Chr.5: 115870132-115870133 on Build GRCh38
rs926063497 Chr.5: 115870177-115870177 on Build GRCh38
rs763732964 Chr.5: 115870130-115870131 on Build GRCh38
rs753748529 Chr.5: 115870089-115870089 on Build GRCh38
rs771596539 Chr.5: 115870161-115870161 on Build GRCh38
rs1051051383 Chr.5: 115870252-115870252 on Build GRCh38
rs757307886 Chr.5: 115870109-115870109 on Build GRCh38
rs933915309 Chr.5: 115870223-115870223 on Build GRCh38
rs913910483 Chr.5: 115870228-115870231 on Build GRCh38
rs921084654 Chr.5: 115870217-115870217 on Build GRCh38
rs774214928 Chr.5: 115870152-115870152 on Build GRCh38
rs747195731 Chr.5: 115870107-115870107 on Build GRCh38
rs768344026 Chr.5: 115870135-115870136 on Build GRCh38
rs946585800 Chr.5: 115870244-115870244 on Build GRCh38
rs770341388 Chr.5: 115870134-115870134 on Build GRCh38
rs936038012 Chr.5: 115870186-115870186 on Build GRCh38
rs761483946 Chr.5: 115870160-115870160 on Build GRCh38
rs754819334 Chr.5: 115870098-115870098 on Build GRCh38
rs796995568 Chr.5: 115870132-115870133 on Build GRCh38
rs778147153 Chr.5: 115870101-115870101 on Build GRCh38
rs745912737 Chr.5: 115870132-115870132 on Build GRCh38
rs780534232 Chr.5: 115870136-115870136 on Build GRCh38
rs749656959 Chr.5: 115870139-115870139 on Build GRCh38
rs993069292 Chr.5: 115870099-115870099 on Build GRCh38
rs528260526 Chr.5: 115870194-115870194 on Build GRCh38

View Full Product Details