Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 32973487-32973716 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APTX
Entrez Gene ID
54840
Gene Name
aprataxin
Gene Aliases
AOA, AOA1, AXA1, EAOH, EOAHA, FHA-HIT
UniGene
Hs.20158
Gene Chromosome Location
Chr.9: 32883872-33025129 on Build GRCh38
Transcripts
NM_001195248.1 NM_001195249.1 NM_001195250.1 NM_001195251.1 NM_001195252.1 NM_001195254.1 NM_175069.2 NM_175073.2 NR_036576.1 NR_036577.1 NR_036578.1 NR_036579.1 XM_006716791.3 XM_006716792.2 XM_011517938.1 XM_011517939.2 XM_017014831.1 XM_017014832.1 XM_017014833.1 XM_017014834.1 XM_017014835.1 XM_017014836.1 XM_017014837.1 XM_017014838.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4373579 c.904A>G p.R302G Chr.9: 32973623-32973623 on Build GRCh38
5777778 c.903T>A p.G301G Chr.9: 32973624-32973624 on Build GRCh38
6842549 c.953G>T p.R318L Chr.9: 32973574-32973574 on Build GRCh38
1598250 c.953G>A p.R318H Chr.9: 32973574-32973574 on Build GRCh38
1598249 c.917G>A p.R306Q Chr.9: 32973610-32973610 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1018257998 Chr.9: 32973628-32973628 on Build GRCh38
rs12001066 Chr.9: 32973598-32973598 on Build GRCh38
rs376734304 Chr.9: 32973569-32973569 on Build GRCh38
rs772656348 Chr.9: 32973610-32973610 on Build GRCh38
rs141493373 Chr.9: 32973556-32973556 on Build GRCh38
rs764599592 Chr.9: 32973624-32973624 on Build GRCh38
rs371292546 Chr.9: 32973574-32973574 on Build GRCh38
rs770727980 Chr.9: 32973600-32973600 on Build GRCh38
rs746541090 Chr.9: 32973611-32973611 on Build GRCh38
rs750406680 Chr.9: 32973641-32973641 on Build GRCh38
rs775949465 Chr.9: 32973516-32973516 on Build GRCh38
rs770357057 Chr.9: 32973612-32973612 on Build GRCh38
rs1005572979 Chr.9: 32973592-32973592 on Build GRCh38
rs776170265 Chr.9: 32973620-32973620 on Build GRCh38
rs1007288114 Chr.9: 32973667-32973667 on Build GRCh38
rs774598658 Chr.9: 32973626-32973626 on Build GRCh38
rs149867156 Chr.9: 32973644-32973644 on Build GRCh38
rs376003639 Chr.9: 32973627-32973627 on Build GRCh38
rs771607486 Chr.9: 32973606-32973606 on Build GRCh38
rs886044599 Chr.9: 32973538-32973538 on Build GRCh38
rs574404091 Chr.9: 32973629-32973629 on Build GRCh38
rs746837337 Chr.9: 32973588-32973588 on Build GRCh38
rs373961930 Chr.9: 32973575-32973575 on Build GRCh38
rs904293109 Chr.9: 32973654-32973654 on Build GRCh38
rs759989689 Chr.9: 32973655-32973655 on Build GRCh38
rs754791634 Chr.9: 32973581-32973581 on Build GRCh38
rs751746847 Chr.9: 32973547-32973547 on Build GRCh38
rs759723787 Chr.9: 32973547-32973548 on Build GRCh38
rs779001961 Chr.9: 32973542-32973542 on Build GRCh38
rs1047315004 Chr.9: 32973578-32973578 on Build GRCh38
rs374053126 Chr.9: 32973567-32973567 on Build GRCh38
rs763358461 Chr.9: 32973622-32973622 on Build GRCh38
rs1057490043 Chr.9: 32973571-32973571 on Build GRCh38
rs200782765 Chr.9: 32973511-32973511 on Build GRCh38
rs763444563 Chr.9: 32973531-32973531 on Build GRCh38
rs747796522 Chr.9: 32973602-32973602 on Build GRCh38
rs753823355 Chr.9: 32973661-32973661 on Build GRCh38
rs938970569 Chr.9: 32973566-32973566 on Build GRCh38
rs754738237 Chr.9: 32973673-32973673 on Build GRCh38
rs757349121 Chr.9: 32973553-32973553 on Build GRCh38

View Full Product Details