Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 155929567-155929804 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTSO
Entrez Gene ID
1519
Gene Name
cathepsin O
Gene Aliases
CTSO1
UniGene
Hs.75262
Gene Chromosome Location
Chr.4: 155924118-155953896 on Build GRCh38
Transcripts
NM_001334.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5038072 c.795A>G p.G265G Chr.4: 155929585-155929585 on Build GRCh38
1052550 c.730A>G p.I244V Chr.4: 155929650-155929650 on Build GRCh38
1309848 c.719C>A p.P240H Chr.4: 155929661-155929661 on Build GRCh38
460662 c.737A>T p.D246V Chr.4: 155929643-155929643 on Build GRCh38
3601308 c.704T>A p.L235H Chr.4: 155929676-155929676 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768388471 Chr.4: 155929660-155929660 on Build GRCh38
rs781599682 Chr.4: 155929755-155929756 on Build GRCh38
rs746755188 Chr.4: 155929750-155929750 on Build GRCh38
rs745839163 Chr.4: 155929729-155929729 on Build GRCh38
rs937155677 Chr.4: 155929761-155929761 on Build GRCh38
rs755264328 Chr.4: 155929746-155929747 on Build GRCh38
rs765781141 Chr.4: 155929680-155929680 on Build GRCh38
rs781325765 Chr.4: 155929728-155929728 on Build GRCh38
rs772011997 Chr.4: 155929731-155929731 on Build GRCh38
rs755170802 Chr.4: 155929611-155929611 on Build GRCh38
rs369555637 Chr.4: 155929722-155929722 on Build GRCh38
rs745811267 Chr.4: 155929628-155929628 on Build GRCh38
rs768439806 Chr.4: 155929757-155929757 on Build GRCh38
rs776307472 Chr.4: 155929662-155929662 on Build GRCh38
rs552008584 Chr.4: 155929616-155929616 on Build GRCh38
rs1042180364 Chr.4: 155929686-155929686 on Build GRCh38
rs964170812 Chr.4: 155929626-155929626 on Build GRCh38
rs751890992 Chr.4: 155929707-155929707 on Build GRCh38
rs537641892 Chr.4: 155929618-155929618 on Build GRCh38
rs371937185 Chr.4: 155929629-155929629 on Build GRCh38
rs150110652 Chr.4: 155929617-155929617 on Build GRCh38
rs778647108 Chr.4: 155929625-155929625 on Build GRCh38
rs202136346 Chr.4: 155929712-155929712 on Build GRCh38
rs779740135 Chr.4: 155929742-155929742 on Build GRCh38
rs796706035 Chr.4: 155929670-155929670 on Build GRCh38
rs746546421 Chr.4: 155929605-155929605 on Build GRCh38
rs144614643 Chr.4: 155929657-155929657 on Build GRCh38
rs750961413 Chr.4: 155929693-155929693 on Build GRCh38
rs1007955798 Chr.4: 155929594-155929594 on Build GRCh38
rs1003448539 Chr.4: 155929775-155929775 on Build GRCh38
rs775329315 Chr.4: 155929641-155929641 on Build GRCh38
rs763504643 Chr.4: 155929699-155929699 on Build GRCh38
rs766703907 Chr.4: 155929705-155929705 on Build GRCh38
rs1054351989 Chr.4: 155929621-155929621 on Build GRCh38
rs536724089 Chr.4: 155929716-155929716 on Build GRCh38
rs142851384 Chr.4: 155929631-155929631 on Build GRCh38
rs756212769 Chr.4: 155929724-155929724 on Build GRCh38
rs761352356 Chr.4: 155929674-155929674 on Build GRCh38
rs138544185 Chr.4: 155929777-155929777 on Build GRCh38
rs932018305 Chr.4: 155929607-155929607 on Build GRCh38
rs913879458 Chr.4: 155929709-155929709 on Build GRCh38
rs925519057 Chr.4: 155929649-155929649 on Build GRCh38

View Full Product Details