Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 95044217-95044482 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCC4
Entrez Gene ID
10257
Gene Name
ATP binding cassette subfamily C member 4
Gene Aliases
MOAT-B, MOATB, MRP4
UniGene
Hs.508423
Gene Chromosome Location
Chr.13: 95019829-95301550 on Build GRCh38
Transcripts
NM_001301829.1 NM_005845.4 XM_005254025.2 XM_017020319.1 XM_017020321.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1368035 c.3566G>T p.R1189M Chr.13: 95044329-95044329 on Build GRCh38
3814149 c.3493A>G p.K1165E Chr.13: 95044402-95044402 on Build GRCh38
7105891 c.3591A>G p.I1197M Chr.13: 95044304-95044304 on Build GRCh38
5020349 c.3609G>A p.A1203A Chr.13: 95044286-95044286 on Build GRCh38
1181343 c.3608C>T p.A1203V Chr.13: 95044287-95044287 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754835483 Chr.13: 95044335-95044335 on Build GRCh38
rs945862790 Chr.13: 95044303-95044303 on Build GRCh38
rs141693430 Chr.13: 95044345-95044345 on Build GRCh38
rs756611814 Chr.13: 95044355-95044355 on Build GRCh38
rs773151365 Chr.13: 95044306-95044306 on Build GRCh38
rs936033278 Chr.13: 95044284-95044284 on Build GRCh38
rs768841551 Chr.13: 95044452-95044454 on Build GRCh38
rs368891562 Chr.13: 95044249-95044249 on Build GRCh38
rs748340394 Chr.13: 95044408-95044408 on Build GRCh38
rs368708883 Chr.13: 95044433-95044433 on Build GRCh38
rs770141439 Chr.13: 95044313-95044313 on Build GRCh38
rs761880861 Chr.13: 95044291-95044291 on Build GRCh38
rs370771661 Chr.13: 95044248-95044248 on Build GRCh38
rs774588723 Chr.13: 95044407-95044407 on Build GRCh38
rs766458578 Chr.13: 95044317-95044317 on Build GRCh38
rs757880054 Chr.13: 95044346-95044346 on Build GRCh38
rs760665091 Chr.13: 95044311-95044311 on Build GRCh38
rs747462604 Chr.13: 95044415-95044416 on Build GRCh38
rs533884311 Chr.13: 95044420-95044420 on Build GRCh38
rs757959172 Chr.13: 95044440-95044440 on Build GRCh38
rs146275324 Chr.13: 95044432-95044432 on Build GRCh38
rs11568695 Chr.13: 95044286-95044286 on Build GRCh38
rs751052644 Chr.13: 95044462-95044462 on Build GRCh38
rs780593353 Chr.13: 95044364-95044364 on Build GRCh38
rs923403997 Chr.13: 95044419-95044419 on Build GRCh38
rs200063919 Chr.13: 95044325-95044325 on Build GRCh38
rs752590278 Chr.13: 95044438-95044438 on Build GRCh38
rs764878414 Chr.13: 95044339-95044339 on Build GRCh38
rs764968059 Chr.13: 95044437-95044437 on Build GRCh38
rs530152342 Chr.13: 95044246-95044246 on Build GRCh38
rs772385215 Chr.13: 95044413-95044413 on Build GRCh38
rs568014366 Chr.13: 95044406-95044406 on Build GRCh38
rs749593762 Chr.13: 95044405-95044405 on Build GRCh38
rs373949612 Chr.13: 95044287-95044287 on Build GRCh38
rs117387249 Chr.13: 95044461-95044461 on Build GRCh38
rs760902252 Chr.13: 95044423-95044423 on Build GRCh38
rs371951064 Chr.13: 95044300-95044300 on Build GRCh38
rs145958534 Chr.13: 95044283-95044283 on Build GRCh38
rs548257801 Chr.13: 95044354-95044354 on Build GRCh38

View Full Product Details