Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 24320111-24320368 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM28
Entrez Gene ID
10863
Gene Name
ADAM metallopeptidase domain 28
Gene Aliases
ADAM 28, MDC-L, MDCL, eMDC II, eMDCII
UniGene
Hs.174030
Gene Chromosome Location
Chr.8: 24294040-24359018 on Build GRCh38
Transcripts
NM_001304351.1 NM_014265.5 NM_021777.4 NR_130709.1 NR_130710.1 XM_005273380.3 XM_005273382.1 XM_006716273.3 XM_006716274.1 XM_011544367.2 XM_011544368.2 XM_011544369.2 XM_011544370.2 XM_017012974.1 XM_017012975.1 XM_017012976.1 NR_125808.1

Gene Symbol
LOC101929294
Entrez Gene ID
101929294
Gene Name
uncharacterized LOC101929294
Gene Aliases
-
UniGene
Hs.637673
Gene Chromosome Location
Chr.8: 24295814-24548618 on Build GRCh38
Transcripts
NM_001304351.1 NM_014265.5 NM_021777.4 NR_130709.1 NR_130710.1 XM_005273380.3 XM_005273382.1 XM_006716273.3 XM_006716274.1 XM_011544367.2 XM_011544368.2 XM_011544369.2 XM_011544370.2 XM_017012974.1 XM_017012975.1 XM_017012976.1 NR_125808.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3736467 c.603A>C p.E201D Chr.8: 24320262-24320262 on Build GRCh38
298571 c.586G>T p.D196Y Chr.8: 24320245-24320245 on Build GRCh38
4572079 c.639T>C p.D213D Chr.8: 24320298-24320298 on Build GRCh38
6399785 c.628T>A p.L210M Chr.8: 24320287-24320287 on Build GRCh38
3736466 c.603A>C p.E201D Chr.8: 24320262-24320262 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745766428 Chr.8: 24320189-24320189 on Build GRCh38
rs939605686 Chr.8: 24320269-24320269 on Build GRCh38
rs530564988 Chr.8: 24320320-24320320 on Build GRCh38
rs781302473 Chr.8: 24320307-24320307 on Build GRCh38
rs773617138 Chr.8: 24320240-24320240 on Build GRCh38
rs563118876 Chr.8: 24320219-24320219 on Build GRCh38
rs758386248 Chr.8: 24320194-24320194 on Build GRCh38
rs976314329 Chr.8: 24320239-24320239 on Build GRCh38
rs371928859 Chr.8: 24320227-24320227 on Build GRCh38
rs756737269 Chr.8: 24320263-24320263 on Build GRCh38
rs749368891 Chr.8: 24320327-24320327 on Build GRCh38
rs763503422 Chr.8: 24320245-24320245 on Build GRCh38
rs746795815 Chr.8: 24320218-24320218 on Build GRCh38
rs754753710 Chr.8: 24320279-24320279 on Build GRCh38
rs915036298 Chr.8: 24320170-24320170 on Build GRCh38
rs771489977 Chr.8: 24320334-24320334 on Build GRCh38
rs985291305 Chr.8: 24320159-24320159 on Build GRCh38
rs13271763 Chr.8: 24320322-24320322 on Build GRCh38
rs190335158 Chr.8: 24320137-24320137 on Build GRCh38
rs1008827189 Chr.8: 24320333-24320333 on Build GRCh38
rs1051114778 Chr.8: 24320315-24320315 on Build GRCh38
rs994545778 Chr.8: 24320305-24320305 on Build GRCh38
rs766844215 Chr.8: 24320253-24320253 on Build GRCh38
rs776855119 Chr.8: 24320223-24320223 on Build GRCh38
rs761543729 Chr.8: 24320150-24320150 on Build GRCh38
rs746814112 Chr.8: 24320276-24320276 on Build GRCh38
rs199883797 Chr.8: 24320262-24320262 on Build GRCh38
rs758476151 Chr.8: 24320274-24320274 on Build GRCh38
rs759885115 Chr.8: 24320256-24320256 on Build GRCh38
rs140515985 Chr.8: 24320206-24320206 on Build GRCh38
rs577876402 Chr.8: 24320167-24320167 on Build GRCh38
rs748183030 Chr.8: 24320226-24320226 on Build GRCh38
rs200839167 Chr.8: 24320275-24320275 on Build GRCh38
rs897266285 Chr.8: 24320302-24320302 on Build GRCh38
rs977816651 Chr.8: 24320238-24320238 on Build GRCh38
rs182572137 Chr.8: 24320330-24320330 on Build GRCh38
rs764848218 Chr.8: 24320266-24320266 on Build GRCh38
rs962502948 Chr.8: 24320180-24320180 on Build GRCh38
rs763653445 Chr.8: 24320260-24320260 on Build GRCh38
rs370669549 Chr.8: 24320235-24320235 on Build GRCh38
rs774767955 Chr.8: 24320254-24320254 on Build GRCh38

View Full Product Details