Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 38458041-38458314 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP23
Entrez Gene ID
57636
Gene Name
Rho GTPase activating protein 23
Gene Aliases
-
UniGene
Hs.374446
Gene Chromosome Location
Chr.17: 38419286-38512392 on Build GRCh38
Transcripts
NM_001199417.1 XM_006721991.1 XM_006721992.3 XM_006721993.3 XM_006721994.3 XM_011525073.1 XM_011525074.2 XM_011525076.2 XM_011525077.2 XM_017024902.1

SNP Detail

SNP ID Chromosome Location
rs866666178 Chr.17: 38458107-38458107 on Build GRCh38
rs935027184 Chr.17: 38458137-38458137 on Build GRCh38
rs774458568 Chr.17: 38458182-38458182 on Build GRCh38
rs1028694681 Chr.17: 38458167-38458167 on Build GRCh38
rs971991014 Chr.17: 38458224-38458224 on Build GRCh38
rs16965349 Chr.17: 38458280-38458280 on Build GRCh38
rs947528692 Chr.17: 38458087-38458087 on Build GRCh38
rs901492671 Chr.17: 38458141-38458141 on Build GRCh38
rs775750536 Chr.17: 38458161-38458161 on Build GRCh38
rs868241187 Chr.17: 38458246-38458246 on Build GRCh38
rs1052560710 Chr.17: 38458138-38458138 on Build GRCh38
rs1005968661 Chr.17: 38458170-38458170 on Build GRCh38
rs548148540 Chr.17: 38458068-38458068 on Build GRCh38
rs574650843 Chr.17: 38458073-38458073 on Build GRCh38
rs768810537 Chr.17: 38458181-38458181 on Build GRCh38
rs968894008 Chr.17: 38458277-38458277 on Build GRCh38
rs377221251 Chr.17: 38458237-38458237 on Build GRCh38
rs142490391 Chr.17: 38458145-38458145 on Build GRCh38
rs186701726 Chr.17: 38458272-38458272 on Build GRCh38
rs761951553 Chr.17: 38458190-38458190 on Build GRCh38
rs540444111 Chr.17: 38458214-38458214 on Build GRCh38
rs746389090 Chr.17: 38458106-38458106 on Build GRCh38
rs757834097 Chr.17: 38458091-38458091 on Build GRCh38
rs151011696 Chr.17: 38458241-38458241 on Build GRCh38
rs776496515 Chr.17: 38458245-38458245 on Build GRCh38
rs961906120 Chr.17: 38458207-38458207 on Build GRCh38
rs377612823 Chr.17: 38458109-38458109 on Build GRCh38
rs781488911 Chr.17: 38458142-38458142 on Build GRCh38
rs568265287 Chr.17: 38458083-38458083 on Build GRCh38
rs931174099 Chr.17: 38458284-38458284 on Build GRCh38
rs866735376 Chr.17: 38458279-38458279 on Build GRCh38
rs370940768 Chr.17: 38458166-38458166 on Build GRCh38
rs781779085 Chr.17: 38458097-38458097 on Build GRCh38
rs888959390 Chr.17: 38458168-38458168 on Build GRCh38
rs958214386 Chr.17: 38458117-38458117 on Build GRCh38
rs746554578 Chr.17: 38458080-38458080 on Build GRCh38
rs977493853 Chr.17: 38458242-38458242 on Build GRCh38

View Full Product Details