Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 125321286-125321535 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAT4
Entrez Gene ID
79633
Gene Name
FAT atypical cadherin 4
Gene Aliases
CDHF14, CDHR11, FAT-J, FATJ, HKLLS2, NBLA00548, VMLDS2
UniGene
Hs.563205
Gene Chromosome Location
Chr.4: 125314913-125492932 on Build GRCh38
Transcripts
NM_001291285.1 NM_001291303.1 NM_024582.4 XM_011532236.2 XM_011532237.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1252165 c.4928C>A p.P1643H Chr.4: 125321339-125321339 on Build GRCh38
5564657 c.5012G>A p.R1671H Chr.4: 125321423-125321423 on Build GRCh38
480821 c.5096A>T p.E1699V Chr.4: 125321507-125321507 on Build GRCh38
1206667 c.4924G>T p.E1642* Chr.4: 125321335-125321335 on Build GRCh38
5709040 c.4958C>T p.A1653V Chr.4: 125321369-125321369 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375605783 Chr.4: 125321423-125321423 on Build GRCh38
rs757579857 Chr.4: 125321392-125321392 on Build GRCh38
rs774671625 Chr.4: 125321356-125321356 on Build GRCh38
rs754645745 Chr.4: 125321409-125321409 on Build GRCh38
rs867783010 Chr.4: 125321482-125321482 on Build GRCh38
rs557542395 Chr.4: 125321445-125321445 on Build GRCh38
rs750795069 Chr.4: 125321487-125321487 on Build GRCh38
rs374190598 Chr.4: 125321490-125321490 on Build GRCh38
rs754225010 Chr.4: 125321381-125321381 on Build GRCh38
rs775639653 Chr.4: 125321372-125321372 on Build GRCh38
rs1047628671 Chr.4: 125321391-125321391 on Build GRCh38
rs777164187 Chr.4: 125321467-125321467 on Build GRCh38
rs201688197 Chr.4: 125321364-125321364 on Build GRCh38
rs369211657 Chr.4: 125321410-125321410 on Build GRCh38
rs777384285 Chr.4: 125321504-125321504 on Build GRCh38
rs746553908 Chr.4: 125321338-125321338 on Build GRCh38
rs376422522 Chr.4: 125321465-125321465 on Build GRCh38
rs765645389 Chr.4: 125321478-125321478 on Build GRCh38
rs201005648 Chr.4: 125321328-125321328 on Build GRCh38
rs572010193 Chr.4: 125321375-125321375 on Build GRCh38
rs374640692 Chr.4: 125321454-125321454 on Build GRCh38
rs747624124 Chr.4: 125321417-125321417 on Build GRCh38
rs539231738 Chr.4: 125321397-125321397 on Build GRCh38
rs555458566 Chr.4: 125321468-125321468 on Build GRCh38
rs1031842031 Chr.4: 125321493-125321493 on Build GRCh38
rs755643759 Chr.4: 125321503-125321503 on Build GRCh38
rs371042493 Chr.4: 125321441-125321441 on Build GRCh38
rs758060257 Chr.4: 125321318-125321318 on Build GRCh38
rs200007224 Chr.4: 125321488-125321488 on Build GRCh38
rs754485366 Chr.4: 125321341-125321341 on Build GRCh38
rs368051619 Chr.4: 125321511-125321511 on Build GRCh38
rs749029819 Chr.4: 125321353-125321353 on Build GRCh38
rs184884584 Chr.4: 125321352-125321352 on Build GRCh38
rs771297547 Chr.4: 125321355-125321355 on Build GRCh38
rs373116363 Chr.4: 125321422-125321422 on Build GRCh38
rs759785439 Chr.4: 125321362-125321362 on Build GRCh38
rs769765728 Chr.4: 125321348-125321348 on Build GRCh38
rs547299465 Chr.4: 125321373-125321373 on Build GRCh38
rs569440986 Chr.4: 125321446-125321446 on Build GRCh38
rs749123036 Chr.4: 125321429-125321429 on Build GRCh38
rs760759561 Chr.4: 125321464-125321464 on Build GRCh38
rs201292031 Chr.4: 125321447-125321447 on Build GRCh38
rs754135078 Chr.4: 125321314-125321314 on Build GRCh38
rs984457402 Chr.4: 125321439-125321439 on Build GRCh38
rs923202662 Chr.4: 125321357-125321357 on Build GRCh38
rs751217207 Chr.4: 125321403-125321403 on Build GRCh38
rs769625100 Chr.4: 125321431-125321432 on Build GRCh38
rs369716271 Chr.4: 125321347-125321347 on Build GRCh38

View Full Product Details