Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 40349847-40350120 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADSL
Entrez Gene ID
158
Gene Name
adenylosuccinate lyase
Gene Aliases
AMPS, ASASE, ASL
UniGene
Hs.75527
Gene Chromosome Location
Chr.22: 40346500-40387408 on Build GRCh38
Transcripts
NM_000026.3 NM_001123378.2 NM_001317923.1 NR_134256.1 XM_011529977.2 XM_011529979.1 XM_011529980.2 XM_017028636.1 XM_017028637.1 XM_017028638.1 XM_017028639.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
445028 c.335C>A p.S112Y Chr.22: 40350013-40350013 on Build GRCh38
5655352 c.329C>A p.A110D Chr.22: 40350007-40350007 on Build GRCh38
6595638 c.247C>T p.R83C Chr.22: 40349925-40349925 on Build GRCh38
5974391 c.259G>C p.D87H Chr.22: 40349937-40349937 on Build GRCh38
1632614 c.256C>A p.H86N Chr.22: 40349934-40349934 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761359362 Chr.22: 40349985-40349985 on Build GRCh38
rs563190965 Chr.22: 40350082-40350082 on Build GRCh38
rs747255804 Chr.22: 40349909-40349909 on Build GRCh38
rs371711909 Chr.22: 40350058-40350058 on Build GRCh38
rs905003988 Chr.22: 40350000-40350000 on Build GRCh38
rs747492297 Chr.22: 40349933-40349933 on Build GRCh38
rs199993991 Chr.22: 40350042-40350042 on Build GRCh38
rs1021686249 Chr.22: 40350044-40350044 on Build GRCh38
rs369139561 Chr.22: 40349983-40349983 on Build GRCh38
rs1035500320 Chr.22: 40350019-40350019 on Build GRCh38
rs147017931 Chr.22: 40350020-40350020 on Build GRCh38
rs763852823 Chr.22: 40349952-40349952 on Build GRCh38
rs559415958 Chr.22: 40350014-40350014 on Build GRCh38
rs752349207 Chr.22: 40349876-40349876 on Build GRCh38
rs145306334 Chr.22: 40349904-40349904 on Build GRCh38
rs763502864 Chr.22: 40350078-40350078 on Build GRCh38
rs181628906 Chr.22: 40350041-40350041 on Build GRCh38
rs867916550 Chr.22: 40350043-40350044 on Build GRCh38
rs752671635 Chr.22: 40350044-40350045 on Build GRCh38
rs974701398 Chr.22: 40350088-40350088 on Build GRCh38
rs758234602 Chr.22: 40350092-40350092 on Build GRCh38
rs897772935 Chr.22: 40349943-40349943 on Build GRCh38
rs149165656 Chr.22: 40349941-40349941 on Build GRCh38
rs200169254 Chr.22: 40349883-40349883 on Build GRCh38
rs374259530 Chr.22: 40350018-40350018 on Build GRCh38
rs796052251 Chr.22: 40349875-40349875 on Build GRCh38
rs750195237 Chr.22: 40350090-40350090 on Build GRCh38
rs1015508318 Chr.22: 40350046-40350046 on Build GRCh38
rs780425464 Chr.22: 40349925-40349925 on Build GRCh38
rs774811008 Chr.22: 40350064-40350064 on Build GRCh38
rs775646855 Chr.22: 40349907-40349907 on Build GRCh38
rs757135310 Chr.22: 40349967-40349967 on Build GRCh38
rs148303726 Chr.22: 40349951-40349951 on Build GRCh38
rs765503061 Chr.22: 40349926-40349926 on Build GRCh38
rs772345709 Chr.22: 40349902-40349902 on Build GRCh38
rs765095853 Chr.22: 40350086-40350086 on Build GRCh38
rs887502738 Chr.22: 40349903-40349903 on Build GRCh38
rs758355156 Chr.22: 40350009-40350009 on Build GRCh38
rs761306504 Chr.22: 40350083-40350083 on Build GRCh38
rs776685935 Chr.22: 40349923-40349923 on Build GRCh38
rs755518176 Chr.22: 40349949-40349949 on Build GRCh38
rs540648461 Chr.22: 40349932-40349932 on Build GRCh38
rs1036185928 Chr.22: 40349931-40349931 on Build GRCh38
rs781534496 Chr.22: 40350029-40350029 on Build GRCh38
rs763470668 Chr.22: 40350057-40350057 on Build GRCh38
rs745787396 Chr.22: 40349895-40349895 on Build GRCh38
rs768958353 Chr.22: 40349922-40349922 on Build GRCh38
rs768596079 Chr.22: 40350025-40350025 on Build GRCh38
rs141408072 Chr.22: 40349960-40349960 on Build GRCh38
rs562234311 Chr.22: 40349891-40349891 on Build GRCh38
rs145786986 Chr.22: 40349894-40349894 on Build GRCh38
rs760230811 Chr.22: 40350070-40350070 on Build GRCh38
rs143275316 Chr.22: 40349948-40349948 on Build GRCh38
rs119450942 Chr.22: 40349976-40349976 on Build GRCh38

View Full Product Details