Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161049345-161049594 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP30
Entrez Gene ID
257106
Gene Name
Rho GTPase activating protein 30
Gene Aliases
-
UniGene
Hs.389374
Gene Chromosome Location
Chr.1: 161046942-161069971 on Build GRCh38
Transcripts
NM_001025598.1 NM_001287600.1 NM_001287602.1 NM_181720.2 XM_005245070.2 XM_005245073.3 XM_011509391.2 XM_017000960.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
898439 c.1547C>T p.S516L Chr.1: 161049563-161049563 on Build GRCh38
5543725 c.1536C>T p.F512F Chr.1: 161049574-161049574 on Build GRCh38
6451113 c.1626C>T p.S542S Chr.1: 161049484-161049484 on Build GRCh38
898441 c.1529_1531delTCT p.F510delF Chr.1: 161049579-161049581 on Build GRCh38
898438 c.1547C>T p.S516L Chr.1: 161049563-161049563 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766699616 Chr.1: 161049439-161049439 on Build GRCh38
rs917881473 Chr.1: 161049510-161049510 on Build GRCh38
rs1029730772 Chr.1: 161049428-161049428 on Build GRCh38
rs373659984 Chr.1: 161049394-161049394 on Build GRCh38
rs758073805 Chr.1: 161049514-161049514 on Build GRCh38
rs769517403 Chr.1: 161049540-161049540 on Build GRCh38
rs756135430 Chr.1: 161049378-161049378 on Build GRCh38
rs756141598 Chr.1: 161049495-161049495 on Build GRCh38
rs574872500 Chr.1: 161049411-161049411 on Build GRCh38
rs757470666 Chr.1: 161049493-161049493 on Build GRCh38
rs761510561 Chr.1: 161049434-161049434 on Build GRCh38
rs375000593 Chr.1: 161049369-161049369 on Build GRCh38
rs759957158 Chr.1: 161049548-161049548 on Build GRCh38
rs745700997 Chr.1: 161049509-161049509 on Build GRCh38
rs989647682 Chr.1: 161049471-161049471 on Build GRCh38
rs993686442 Chr.1: 161049400-161049400 on Build GRCh38
rs746534176 Chr.1: 161049419-161049419 on Build GRCh38
rs759778089 Chr.1: 161049455-161049455 on Build GRCh38
rs183851032 Chr.1: 161049549-161049549 on Build GRCh38
rs775522404 Chr.1: 161049418-161049418 on Build GRCh38
rs780823026 Chr.1: 161049536-161049536 on Build GRCh38
rs569309380 Chr.1: 161049472-161049472 on Build GRCh38
rs768032737 Chr.1: 161049459-161049459 on Build GRCh38
rs774746220 Chr.1: 161049446-161049446 on Build GRCh38
rs757060599 Chr.1: 161049409-161049409 on Build GRCh38
rs191146080 Chr.1: 161049412-161049412 on Build GRCh38
rs145240665 Chr.1: 161049569-161049569 on Build GRCh38
rs201650598 Chr.1: 161049519-161049519 on Build GRCh38
rs981792070 Chr.1: 161049429-161049429 on Build GRCh38
rs749509929 Chr.1: 161049396-161049396 on Build GRCh38
rs756255893 Chr.1: 161049482-161049482 on Build GRCh38
rs768212801 Chr.1: 161049424-161049424 on Build GRCh38
rs746936756 Chr.1: 161049520-161049520 on Build GRCh38
rs150017851 Chr.1: 161049495-161049495 on Build GRCh38
rs776037650 Chr.1: 161049561-161049561 on Build GRCh38
rs776490199 Chr.1: 161049431-161049431 on Build GRCh38
rs754110109 Chr.1: 161049490-161049490 on Build GRCh38
rs764147345 Chr.1: 161049483-161049483 on Build GRCh38
rs959642210 Chr.1: 161049442-161049442 on Build GRCh38
rs1020800832 Chr.1: 161049460-161049460 on Build GRCh38
rs200537305 Chr.1: 161049388-161049388 on Build GRCh38
rs747792122 Chr.1: 161049538-161049538 on Build GRCh38
rs772691068 Chr.1: 161049547-161049547 on Build GRCh38
rs768335781 Chr.1: 161049533-161049533 on Build GRCh38
rs1039321893 Chr.1: 161049390-161049390 on Build GRCh38

View Full Product Details