Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 9211409-9211652 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD12
Entrez Gene ID
23253
Gene Name
ankyrin repeat domain 12
Gene Aliases
ANCO-2, ANCO1, GAC-1, Nbla00144
UniGene
Hs.464585
Gene Chromosome Location
Chr.18: 9136753-9285985 on Build GRCh38
Transcripts
NM_001083625.2 NM_001204056.1 NM_015208.4 XM_005258092.3 XM_005258093.3 XM_005258095.2 XM_005258096.2 XM_011525638.2 XM_011525639.2 XM_017025661.1 XM_017025662.1 XM_017025663.1 XM_017025664.1 XM_017025665.1 XM_017025666.1 XM_017025667.1 XM_017025668.1 XM_017025670.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5544477 c.470C>T p.P157L Chr.18: 9211602-9211602 on Build GRCh38
474079 c.468T>C p.H156H Chr.18: 9211600-9211600 on Build GRCh38
4140194 c.483G>C p.T161T Chr.18: 9211615-9211615 on Build GRCh38
438423 c.482C>T p.T161M Chr.18: 9211614-9211614 on Build GRCh38
268661 c.483G>A p.T161T Chr.18: 9211615-9211615 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs547526796 Chr.18: 9211472-9211472 on Build GRCh38
rs770219739 Chr.18: 9211548-9211548 on Build GRCh38
rs750707861 Chr.18: 9211588-9211588 on Build GRCh38
rs901917085 Chr.18: 9211487-9211487 on Build GRCh38
rs56105700 Chr.18: 9211478-9211478 on Build GRCh38
rs754679529 Chr.18: 9211589-9211589 on Build GRCh38
rs746197769 Chr.18: 9211537-9211537 on Build GRCh38
rs752509973 Chr.18: 9211597-9211597 on Build GRCh38
rs1024039539 Chr.18: 9211532-9211532 on Build GRCh38
rs367715555 Chr.18: 9211556-9211556 on Build GRCh38
rs780779667 Chr.18: 9211614-9211614 on Build GRCh38
rs775805789 Chr.18: 9211550-9211550 on Build GRCh38
rs902486791 Chr.18: 9211439-9211439 on Build GRCh38
rs993015993 Chr.18: 9211514-9211514 on Build GRCh38
rs1015178827 Chr.18: 9211587-9211587 on Build GRCh38
rs765091845 Chr.18: 9211593-9211593 on Build GRCh38
rs988620952 Chr.18: 9211616-9211616 on Build GRCh38
rs548984879 Chr.18: 9211486-9211489 on Build GRCh38
rs749674116 Chr.18: 9211627-9211627 on Build GRCh38
rs764580080 Chr.18: 9211568-9211568 on Build GRCh38
rs371533373 Chr.18: 9211615-9211615 on Build GRCh38
rs756797426 Chr.18: 9211611-9211611 on Build GRCh38
rs979730008 Chr.18: 9211551-9211551 on Build GRCh38
rs776312572 Chr.18: 9211535-9211535 on Build GRCh38
rs758351899 Chr.18: 9211600-9211600 on Build GRCh38
rs767819715 Chr.18: 9211577-9211577 on Build GRCh38
rs746496404 Chr.18: 9211609-9211609 on Build GRCh38
rs751122885 Chr.18: 9211571-9211574 on Build GRCh38
rs950509684 Chr.18: 9211503-9211503 on Build GRCh38
rs777742137 Chr.18: 9211603-9211603 on Build GRCh38
rs767707285 Chr.18: 9211574-9211574 on Build GRCh38
rs775921994 Chr.18: 9211625-9211625 on Build GRCh38
rs1040474815 Chr.18: 9211485-9211485 on Build GRCh38
rs995007256 Chr.18: 9211460-9211460 on Build GRCh38
rs530922119 Chr.18: 9211491-9211491 on Build GRCh38
rs766157386 Chr.18: 9211566-9211567 on Build GRCh38

View Full Product Details