Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 57482309-57482540 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTBL2
Entrez Gene ID
345651
Gene Name
actin, beta like 2
Gene Aliases
ACT
UniGene
Hs.482167
Gene Chromosome Location
Chr.5: 57480016-57482809 on Build GRCh38
Transcripts
NM_001017992.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
738466 c.389C>A p.T130K Chr.5: 57482319-57482319 on Build GRCh38
4423283 c.172G>A p.E58K Chr.5: 57482536-57482536 on Build GRCh38
1132133 c.207G>T p.K69N Chr.5: 57482501-57482501 on Build GRCh38
1069091 c.244G>A p.D82N Chr.5: 57482464-57482464 on Build GRCh38
3410334 c.217G>A p.E73K Chr.5: 57482491-57482491 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777902137 Chr.5: 57482332-57482332 on Build GRCh38
rs560370286 Chr.5: 57482516-57482516 on Build GRCh38
rs891844089 Chr.5: 57482434-57482434 on Build GRCh38
rs199915773 Chr.5: 57482340-57482340 on Build GRCh38
rs373420374 Chr.5: 57482405-57482405 on Build GRCh38
rs905848404 Chr.5: 57482413-57482413 on Build GRCh38
rs371334458 Chr.5: 57482502-57482502 on Build GRCh38
rs769733989 Chr.5: 57482464-57482464 on Build GRCh38
rs763928355 Chr.5: 57482468-57482468 on Build GRCh38
rs150453337 Chr.5: 57482469-57482469 on Build GRCh38
rs753169150 Chr.5: 57482337-57482337 on Build GRCh38
rs371146209 Chr.5: 57482387-57482387 on Build GRCh38
rs140516050 Chr.5: 57482460-57482460 on Build GRCh38
rs148214432 Chr.5: 57482478-57482478 on Build GRCh38
rs1029757499 Chr.5: 57482462-57482462 on Build GRCh38
rs781191771 Chr.5: 57482401-57482401 on Build GRCh38
rs745849436 Chr.5: 57482356-57482356 on Build GRCh38
rs73757391 Chr.5: 57482386-57482386 on Build GRCh38
rs201074919 Chr.5: 57482498-57482498 on Build GRCh38
rs780632687 Chr.5: 57482358-57482358 on Build GRCh38
rs755518154 Chr.5: 57482510-57482510 on Build GRCh38
rs756215581 Chr.5: 57482491-57482491 on Build GRCh38
rs145587699 Chr.5: 57482374-57482374 on Build GRCh38
rs142323625 Chr.5: 57482404-57482404 on Build GRCh38
rs76155491 Chr.5: 57482376-57482376 on Build GRCh38
rs759588880 Chr.5: 57482339-57482339 on Build GRCh38
rs375975550 Chr.5: 57482508-57482508 on Build GRCh38
rs371341145 Chr.5: 57482470-57482470 on Build GRCh38
rs781303825 Chr.5: 57482345-57482345 on Build GRCh38
rs200674727 Chr.5: 57482365-57482365 on Build GRCh38
rs1000719305 Chr.5: 57482422-57482422 on Build GRCh38
rs147643155 Chr.5: 57482398-57482398 on Build GRCh38
rs776182541 Chr.5: 57482385-57482385 on Build GRCh38
rs775355094 Chr.5: 57482465-57482465 on Build GRCh38
rs749694409 Chr.5: 57482407-57482407 on Build GRCh38
rs771865618 Chr.5: 57482360-57482360 on Build GRCh38
rs762676574 Chr.5: 57482389-57482389 on Build GRCh38
rs757482999 Chr.5: 57482400-57482400 on Build GRCh38
rs149617159 Chr.5: 57482486-57482486 on Build GRCh38
rs139117678 Chr.5: 57482439-57482439 on Build GRCh38
rs746415066 Chr.5: 57482354-57482354 on Build GRCh38
rs780739257 Chr.5: 57482492-57482492 on Build GRCh38
rs987715830 Chr.5: 57482421-57482421 on Build GRCh38
rs200839938 Chr.5: 57482359-57482359 on Build GRCh38
rs747123764 Chr.5: 57482335-57482335 on Build GRCh38
rs776067447 Chr.5: 57482447-57482447 on Build GRCh38
rs770414529 Chr.5: 57482357-57482357 on Build GRCh38
rs764333556 Chr.5: 57482395-57482395 on Build GRCh38
rs779203289 Chr.5: 57482515-57482515 on Build GRCh38
rs772952110 Chr.5: 57482363-57482363 on Build GRCh38
rs762848446 Chr.5: 57482467-57482467 on Build GRCh38
rs147178651 Chr.5: 57482408-57482408 on Build GRCh38
rs779044293 Chr.5: 57482330-57482341 on Build GRCh38
rs746689027 Chr.5: 57482427-57482427 on Build GRCh38

View Full Product Details