Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 91587855-91588080 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CATSPERB
Entrez Gene ID
79820
Gene Name
cation channel sperm associated auxiliary subunit beta
Gene Aliases
C14orf161, CatSper(beta)
UniGene
Hs.131755
Gene Chromosome Location
Chr.14: 91580774-91732086 on Build GRCh38
Transcripts
NM_024764.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
106995 c.2988G>A p.M996I Chr.14: 91588047-91588047 on Build GRCh38
4655662 c.2983A>C p.R995R Chr.14: 91588052-91588052 on Build GRCh38
4453478 c.3007A>G p.I1003V Chr.14: 91588028-91588028 on Build GRCh38
279589 c.3017C>T p.A1006V Chr.14: 91588018-91588018 on Build GRCh38
1199704 c.3010C>A p.L1004I Chr.14: 91588025-91588025 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372921289 Chr.14: 91587975-91587975 on Build GRCh38
rs760005876 Chr.14: 91587951-91587951 on Build GRCh38
rs757857394 Chr.14: 91588019-91588019 on Build GRCh38
rs150042994 Chr.14: 91587889-91587889 on Build GRCh38
rs775458477 Chr.14: 91587953-91587953 on Build GRCh38
rs1013904176 Chr.14: 91587907-91587907 on Build GRCh38
rs140824630 Chr.14: 91588047-91588047 on Build GRCh38
rs1032056386 Chr.14: 91588014-91588015 on Build GRCh38
rs933785041 Chr.14: 91588008-91588008 on Build GRCh38
rs747337132 Chr.14: 91587929-91587929 on Build GRCh38
rs112534589 Chr.14: 91587945-91587945 on Build GRCh38
rs754993660 Chr.14: 91588002-91588002 on Build GRCh38
rs982758168 Chr.14: 91587919-91587919 on Build GRCh38
rs539929563 Chr.14: 91587937-91587937 on Build GRCh38
rs780855784 Chr.14: 91588005-91588007 on Build GRCh38
rs777354519 Chr.14: 91588030-91588030 on Build GRCh38
rs770529626 Chr.14: 91587897-91587897 on Build GRCh38
rs902174087 Chr.14: 91587925-91587925 on Build GRCh38
rs763035368 Chr.14: 91587968-91587968 on Build GRCh38
rs766252095 Chr.14: 91587996-91587996 on Build GRCh38
rs781349604 Chr.14: 91587990-91587990 on Build GRCh38
rs753863516 Chr.14: 91587997-91587997 on Build GRCh38
rs764230511 Chr.14: 91587969-91587969 on Build GRCh38
rs778682545 Chr.14: 91588028-91588028 on Build GRCh38
rs541713211 Chr.14: 91587912-91587912 on Build GRCh38
rs369065234 Chr.14: 91587936-91587936 on Build GRCh38
rs747625080 Chr.14: 91588009-91588009 on Build GRCh38
rs778860336 Chr.14: 91588007-91588007 on Build GRCh38
rs45607735 Chr.14: 91587978-91587978 on Build GRCh38

View Full Product Details