Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 70674575-70674832 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTS14
Entrez Gene ID
140766
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 14
Gene Aliases
-
UniGene
Hs.352156
Gene Chromosome Location
Chr.10: 70672518-70762441 on Build GRCh38
Transcripts
NM_080722.4 NM_139155.3 XM_011539300.2 XM_011539303.2 XM_011539305.2 XM_011539306.2 XM_011539307.2 XM_011539308.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7354272 c.235C>T p.H79Y Chr.10: 70674708-70674708 on Build GRCh38
5663599 c.277G>T p.G93* Chr.10: 70674750-70674750 on Build GRCh38
6517980 c.359G>A p.R120H Chr.10: 70674832-70674832 on Build GRCh38
5803505 c.285C>A p.T95T Chr.10: 70674758-70674758 on Build GRCh38
3440123 c.249C>T p.L83L Chr.10: 70674722-70674722 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777004859 Chr.10: 70674785-70674785 on Build GRCh38
rs138392873 Chr.10: 70674600-70674600 on Build GRCh38
rs148384658 Chr.10: 70674638-70674638 on Build GRCh38
rs751798747 Chr.10: 70674632-70674632 on Build GRCh38
rs757990147 Chr.10: 70674736-70674736 on Build GRCh38
rs768301923 Chr.10: 70674750-70674750 on Build GRCh38
rs757796491 Chr.10: 70674732-70674732 on Build GRCh38
rs748048717 Chr.10: 70674759-70674759 on Build GRCh38
rs371895859 Chr.10: 70674780-70674780 on Build GRCh38
rs201484487 Chr.10: 70674619-70674619 on Build GRCh38
rs538508973 Chr.10: 70674705-70674705 on Build GRCh38
rs760936209 Chr.10: 70674722-70674722 on Build GRCh38
rs753031516 Chr.10: 70674720-70674720 on Build GRCh38
rs936513162 Chr.10: 70674686-70674686 on Build GRCh38
rs990843726 Chr.10: 70674604-70674604 on Build GRCh38
rs141561275 Chr.10: 70674639-70674639 on Build GRCh38
rs771093112 Chr.10: 70674704-70674704 on Build GRCh38
rs745484632 Chr.10: 70674676-70674676 on Build GRCh38
rs769866318 Chr.10: 70674762-70674762 on Build GRCh38
rs144227111 Chr.10: 70674635-70674635 on Build GRCh38
rs1026831167 Chr.10: 70674774-70674774 on Build GRCh38
rs771648076 Chr.10: 70674681-70674681 on Build GRCh38
rs774505808 Chr.10: 70674765-70674765 on Build GRCh38
rs114392999 Chr.10: 70674618-70674618 on Build GRCh38
rs372200693 Chr.10: 70674770-70674770 on Build GRCh38
rs756739834 Chr.10: 70674668-70674668 on Build GRCh38
rs766612764 Chr.10: 70674625-70674625 on Build GRCh38
rs370245201 Chr.10: 70674746-70674746 on Build GRCh38
rs746771070 Chr.10: 70674684-70674684 on Build GRCh38
rs776686112 Chr.10: 70674693-70674693 on Build GRCh38
rs554496606 Chr.10: 70674688-70674688 on Build GRCh38
rs760615488 Chr.10: 70674610-70674610 on Build GRCh38
rs201449180 Chr.10: 70674706-70674706 on Build GRCh38
rs760999643 Chr.10: 70674778-70674778 on Build GRCh38
rs1049554519 Chr.10: 70674689-70674689 on Build GRCh38
rs983420240 Chr.10: 70674789-70674789 on Build GRCh38
rs772253184 Chr.10: 70674773-70674773 on Build GRCh38
rs764571352 Chr.10: 70674723-70674723 on Build GRCh38
rs186907048 Chr.10: 70674813-70674813 on Build GRCh38
rs752860571 Chr.10: 70674652-70674654 on Build GRCh38
rs752458774 Chr.10: 70674737-70674737 on Build GRCh38
rs145982854 Chr.10: 70674751-70674751 on Build GRCh38
rs779844010 Chr.10: 70674683-70674683 on Build GRCh38
rs374197954 Chr.10: 70674624-70674624 on Build GRCh38
rs61754837 Chr.10: 70674733-70674733 on Build GRCh38
rs749952998 Chr.10: 70674734-70674734 on Build GRCh38
rs775554366 Chr.10: 70674775-70674775 on Build GRCh38
rs74141809 Chr.10: 70674708-70674708 on Build GRCh38
rs754601062 Chr.10: 70674649-70674653 on Build GRCh38
rs756578095 Chr.10: 70674642-70674642 on Build GRCh38
rs778438003 Chr.10: 70674675-70674675 on Build GRCh38
rs139657777 Chr.10: 70674665-70674665 on Build GRCh38
rs754314763 Chr.10: 70674724-70674724 on Build GRCh38
rs773117486 Chr.10: 70674812-70674813 on Build GRCh38
rs761881397 Chr.10: 70674694-70674694 on Build GRCh38
rs778768541 Chr.10: 70674807-70674808 on Build GRCh38
rs139509859 Chr.10: 70674812-70674812 on Build GRCh38
rs146369407 Chr.10: 70674658-70674658 on Build GRCh38
rs368175978 Chr.10: 70674757-70674757 on Build GRCh38

View Full Product Details