Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 7676967-7677224 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MCEMP1
Entrez Gene ID
199675
Gene Name
mast cell expressed membrane protein 1
Gene Aliases
C19orf59
UniGene
Hs.709539
Gene Chromosome Location
Chr.19: 7677057-7679833 on Build GRCh38
Transcripts
NM_174918.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5391143 c.4G>A p.E2K Chr.19: 7677085-7677085 on Build GRCh38
5426797 c.35T>C p.V12A Chr.19: 7677116-7677116 on Build GRCh38
6880398 c.74G>A p.G25E Chr.19: 7677155-7677155 on Build GRCh38
399005 c.2T>C p.M1T Chr.19: 7677083-7677083 on Build GRCh38
6990467 c.91C>G p.Q31E Chr.19: 7677172-7677172 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs755517971 Chr.19: 7677188-7677188 on Build GRCh38
rs752045527 Chr.19: 7677051-7677051 on Build GRCh38
rs763241664 Chr.19: 7677175-7677175 on Build GRCh38
rs576910434 Chr.19: 7677156-7677156 on Build GRCh38
rs758637734 Chr.19: 7677039-7677039 on Build GRCh38
rs554922384 Chr.19: 7677007-7677007 on Build GRCh38
rs781775073 Chr.19: 7677190-7677190 on Build GRCh38
rs1009137526 Chr.19: 7677159-7677159 on Build GRCh38
rs753279169 Chr.19: 7677202-7677202 on Build GRCh38
rs539857036 Chr.19: 7677045-7677045 on Build GRCh38
rs538168059 Chr.19: 7676999-7676999 on Build GRCh38
rs747708797 Chr.19: 7677066-7677066 on Build GRCh38
rs750549401 Chr.19: 7677165-7677165 on Build GRCh38
rs373960571 Chr.19: 7677056-7677056 on Build GRCh38
rs866667434 Chr.19: 7677161-7677161 on Build GRCh38
rs748993635 Chr.19: 7677083-7677083 on Build GRCh38
rs775991095 Chr.19: 7677046-7677046 on Build GRCh38
rs574339606 Chr.19: 7677011-7677011 on Build GRCh38
rs759468393 Chr.19: 7677114-7677114 on Build GRCh38
rs904203157 Chr.19: 7677091-7677091 on Build GRCh38
rs766845007 Chr.19: 7677181-7677181 on Build GRCh38
rs999921739 Chr.19: 7677093-7677093 on Build GRCh38
rs144064460 Chr.19: 7677148-7677148 on Build GRCh38
rs939503551 Chr.19: 7677010-7677010 on Build GRCh38
rs200957710 Chr.19: 7677144-7677144 on Build GRCh38
rs769458405 Chr.19: 7677069-7677069 on Build GRCh38
rs181773151 Chr.19: 7676989-7676989 on Build GRCh38
rs770574712 Chr.19: 7677087-7677087 on Build GRCh38
rs375705250 Chr.19: 7677052-7677052 on Build GRCh38
rs1053912349 Chr.19: 7676991-7676991 on Build GRCh38
rs752026943 Chr.19: 7677187-7677187 on Build GRCh38
rs774140999 Chr.19: 7677100-7677100 on Build GRCh38
rs907978270 Chr.19: 7677000-7677000 on Build GRCh38

View Full Product Details