Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 104784246-104784507 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA1
Entrez Gene ID
19
Gene Name
ATP binding cassette subfamily A member 1
Gene Aliases
ABC-1, ABC1, CERP, HDLDT1, TGD
UniGene
Hs.659274
Gene Chromosome Location
Chr.9: 104781002-104928246 on Build GRCh38
Transcripts
NM_005502.3 XM_005251773.2 XM_005251776.2 XM_011518339.2 XM_011518340.2 XM_011518341.2 XM_011518342.2 XM_017014378.1 XM_017014379.1 XM_017014380.1 XM_017014381.1 XM_017014382.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4504767 c.6746C>G p.S2249C Chr.9: 104784355-104784355 on Build GRCh38
3432730 c.6694G>T p.D2232Y Chr.9: 104784407-104784407 on Build GRCh38
1459098 c.6779A>G p.Y2260C Chr.9: 104784322-104784322 on Build GRCh38
4805391 c.6677A>T p.D2226V Chr.9: 104784424-104784424 on Build GRCh38
3432729 c.6712A>C p.N2238H Chr.9: 104784389-104784389 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779593454 Chr.9: 104784298-104784298 on Build GRCh38
rs748538360 Chr.9: 104784329-104784329 on Build GRCh38
rs780958155 Chr.9: 104784321-104784321 on Build GRCh38
rs923910019 Chr.9: 104784419-104784419 on Build GRCh38
rs746020160 Chr.9: 104784357-104784357 on Build GRCh38
rs777670089 Chr.9: 104784449-104784449 on Build GRCh38
rs772446983 Chr.9: 104784358-104784358 on Build GRCh38
rs764486276 Chr.9: 104784273-104784273 on Build GRCh38
rs370983511 Chr.9: 104784293-104784293 on Build GRCh38
rs749070718 Chr.9: 104784471-104784471 on Build GRCh38
rs144588452 Chr.9: 104784371-104784371 on Build GRCh38
rs575454176 Chr.9: 104784289-104784289 on Build GRCh38
rs34879708 Chr.9: 104784372-104784372 on Build GRCh38
rs775831681 Chr.9: 104784366-104784366 on Build GRCh38
rs751049570 Chr.9: 104784318-104784318 on Build GRCh38
rs762540648 Chr.9: 104784400-104784400 on Build GRCh38
rs972456579 Chr.9: 104784304-104784304 on Build GRCh38
rs765921869 Chr.9: 104784406-104784406 on Build GRCh38
rs745893578 Chr.9: 104784341-104784341 on Build GRCh38
rs368920893 Chr.9: 104784453-104784453 on Build GRCh38
rs375241409 Chr.9: 104784295-104784295 on Build GRCh38
rs771743951 Chr.9: 104784414-104784414 on Build GRCh38
rs773601853 Chr.9: 104784396-104784396 on Build GRCh38
rs961308213 Chr.9: 104784302-104784302 on Build GRCh38
rs192904467 Chr.9: 104784333-104784333 on Build GRCh38
rs752621830 Chr.9: 104784421-104784421 on Build GRCh38
rs777237083 Chr.9: 104784275-104784275 on Build GRCh38
rs779300353 Chr.9: 104784306-104784306 on Build GRCh38
rs557180532 Chr.9: 104784283-104784283 on Build GRCh38
rs762785983 Chr.9: 104784299-104784299 on Build GRCh38
rs755958919 Chr.9: 104784348-104784348 on Build GRCh38
rs757115183 Chr.9: 104784477-104784477 on Build GRCh38
rs767111293 Chr.9: 104784417-104784417 on Build GRCh38
rs375203349 Chr.9: 104784354-104784354 on Build GRCh38
rs762324071 Chr.9: 104784380-104784380 on Build GRCh38
rs372148712 Chr.9: 104784299-104784299 on Build GRCh38
rs374793716 Chr.9: 104784447-104784447 on Build GRCh38
rs1039038676 Chr.9: 104784382-104784382 on Build GRCh38
rs371654844 Chr.9: 104784387-104784387 on Build GRCh38
rs765522688 Chr.9: 104784386-104784386 on Build GRCh38
rs368288959 Chr.9: 104784296-104784296 on Build GRCh38
rs751261755 Chr.9: 104784412-104784412 on Build GRCh38

View Full Product Details