Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 218164855-218165116 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXCR1
Entrez Gene ID
3577
Gene Name
C-X-C motif chemokine receptor 1
Gene Aliases
C-C, C-C-CKR-1, CD128, CD181, CDw128a, CKR-1, CMKAR1, IL8R1, IL8RA, IL8RBA
UniGene
Hs.194778
Gene Chromosome Location
Chr.2: 218162845-218166993 on Build GRCh38
Transcripts
NM_000634.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3983917 c.280A>G p.I94V Chr.2: 218164932-218164932 on Build GRCh38
3962391 c.97G>A p.E33K Chr.2: 218165115-218165115 on Build GRCh38
3909590 c.325C>T p.L109L Chr.2: 218164887-218164887 on Build GRCh38
720008 c.208G>T p.G70C Chr.2: 218165004-218165004 on Build GRCh38
6900148 c.159C>A p.S53R Chr.2: 218165053-218165053 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749978322 Chr.2: 218164933-218164933 on Build GRCh38
rs771039240 Chr.2: 218165075-218165075 on Build GRCh38
rs762870290 Chr.2: 218164997-218164997 on Build GRCh38
rs748337900 Chr.2: 218164982-218164982 on Build GRCh38
rs773237679 Chr.2: 218164989-218164989 on Build GRCh38
rs754364462 Chr.2: 218165029-218165029 on Build GRCh38
rs779315622 Chr.2: 218164969-218164969 on Build GRCh38
rs762820875 Chr.2: 218164897-218164897 on Build GRCh38
rs748427779 Chr.2: 218164885-218164885 on Build GRCh38
rs761511554 Chr.2: 218165005-218165005 on Build GRCh38
rs773319684 Chr.2: 218164888-218164888 on Build GRCh38
rs749231087 Chr.2: 218165080-218165080 on Build GRCh38
rs201314907 Chr.2: 218164960-218164960 on Build GRCh38
rs922958326 Chr.2: 218165039-218165039 on Build GRCh38
rs1050426268 Chr.2: 218164936-218164936 on Build GRCh38
rs889982682 Chr.2: 218164950-218164950 on Build GRCh38
rs779225479 Chr.2: 218165043-218165043 on Build GRCh38
rs1008384006 Chr.2: 218164951-218164951 on Build GRCh38
rs774641996 Chr.2: 218164880-218164880 on Build GRCh38
rs765860636 Chr.2: 218165027-218165027 on Build GRCh38
rs767307017 Chr.2: 218165012-218165012 on Build GRCh38
rs113747232 Chr.2: 218164959-218164959 on Build GRCh38
rs774250451 Chr.2: 218165086-218165086 on Build GRCh38
rs758551408 Chr.2: 218165063-218165063 on Build GRCh38
rs538831033 Chr.2: 218165016-218165016 on Build GRCh38
rs547150987 Chr.2: 218165025-218165025 on Build GRCh38
rs1025632169 Chr.2: 218164944-218164944 on Build GRCh38
rs751256472 Chr.2: 218164917-218164917 on Build GRCh38
rs1050375380 Chr.2: 218164882-218164882 on Build GRCh38
rs759445614 Chr.2: 218164892-218164892 on Build GRCh38
rs753081180 Chr.2: 218165059-218165059 on Build GRCh38
rs780611509 Chr.2: 218164940-218164940 on Build GRCh38
rs531740469 Chr.2: 218164935-218164935 on Build GRCh38
rs5021153 Chr.2: 218165035-218165035 on Build GRCh38
rs940418262 Chr.2: 218164923-218164923 on Build GRCh38
rs374045826 Chr.2: 218165079-218165079 on Build GRCh38
rs191545072 Chr.2: 218164905-218164905 on Build GRCh38
rs147633483 Chr.2: 218164995-218164995 on Build GRCh38
rs149163975 Chr.2: 218165004-218165004 on Build GRCh38
rs145296974 Chr.2: 218164928-218164928 on Build GRCh38
rs147031882 Chr.2: 218165000-218165000 on Build GRCh38
rs560768385 Chr.2: 218164886-218164886 on Build GRCh38
rs1805038 Chr.2: 218165001-218165001 on Build GRCh38
rs748986282 Chr.2: 218165079-218165080 on Build GRCh38
rs768739760 Chr.2: 218165083-218165083 on Build GRCh38
rs898598822 Chr.2: 218164926-218164926 on Build GRCh38
rs765253110 Chr.2: 218164893-218164893 on Build GRCh38
rs3092972 Chr.2: 218164924-218164924 on Build GRCh38
rs747253443 Chr.2: 218165074-218165074 on Build GRCh38
rs200296997 Chr.2: 218164901-218164901 on Build GRCh38
rs755356236 Chr.2: 218164967-218164967 on Build GRCh38
rs931614779 Chr.2: 218164934-218164934 on Build GRCh38
rs747117990 Chr.2: 218164991-218164991 on Build GRCh38
rs772327936 Chr.2: 218164986-218164986 on Build GRCh38
rs550231951 Chr.2: 218164996-218164996 on Build GRCh38
rs777985863 Chr.2: 218165066-218165066 on Build GRCh38

View Full Product Details