Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 41609788-41610049 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPAT4
Entrez Gene ID
137964
Gene Name
glycerol-3-phosphate acyltransferase 4
Gene Aliases
1-AGPAT 6, AGPAT6, LPAAT-zeta, LPAATZ, TSARG7
UniGene
Hs.355753
Gene Chromosome Location
Chr.8: 41578188-41625001 on Build GRCh38
Transcripts
NM_178819.3 XM_011544392.2 XM_011544393.1 XM_011544394.1 XM_017013042.1 XM_017013043.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6596403 c.395C>T p.T132I Chr.8: 41609814-41609814 on Build GRCh38
5041242 c.449A>G p.N150S Chr.8: 41609868-41609868 on Build GRCh38
3649163 c.370G>T p.E124* Chr.8: 41609789-41609789 on Build GRCh38
1457065 c.441C>T p.S147S Chr.8: 41609860-41609860 on Build GRCh38
3900159 c.429G>A p.W143* Chr.8: 41609848-41609848 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774380443 Chr.8: 41609855-41609855 on Build GRCh38
rs753407198 Chr.8: 41609896-41609896 on Build GRCh38
rs748281173 Chr.8: 41609962-41609962 on Build GRCh38
rs776375117 Chr.8: 41609994-41609994 on Build GRCh38
rs751826880 Chr.8: 41609923-41609923 on Build GRCh38
rs758917144 Chr.8: 41609913-41609913 on Build GRCh38
rs772068375 Chr.8: 41609976-41609976 on Build GRCh38
rs749360599 Chr.8: 41609973-41609973 on Build GRCh38
rs767316710 Chr.8: 41609875-41609875 on Build GRCh38
rs376842648 Chr.8: 41609906-41609906 on Build GRCh38
rs202065151 Chr.8: 41609851-41609851 on Build GRCh38
rs139697712 Chr.8: 41609908-41609908 on Build GRCh38
rs752549936 Chr.8: 41609880-41609880 on Build GRCh38
rs971058322 Chr.8: 41609921-41609921 on Build GRCh38
rs760508617 Chr.8: 41609987-41609987 on Build GRCh38
rs200325338 Chr.8: 41610000-41610000 on Build GRCh38
rs781500138 Chr.8: 41609954-41609954 on Build GRCh38
rs201136934 Chr.8: 41609902-41609902 on Build GRCh38
rs368282604 Chr.8: 41609945-41609945 on Build GRCh38
rs578249326 Chr.8: 41609839-41609839 on Build GRCh38
rs1002596164 Chr.8: 41609930-41609930 on Build GRCh38
rs762003163 Chr.8: 41609843-41609843 on Build GRCh38
rs763745101 Chr.8: 41609991-41609991 on Build GRCh38
rs914481282 Chr.8: 41609998-41609998 on Build GRCh38
rs747247483 Chr.8: 41609830-41609830 on Build GRCh38
rs759404870 Chr.8: 41609865-41609865 on Build GRCh38
rs776686044 Chr.8: 41609840-41609840 on Build GRCh38
rs201102148 Chr.8: 41609971-41609971 on Build GRCh38
rs199941673 Chr.8: 41609950-41609950 on Build GRCh38
rs267601921 Chr.8: 41609879-41609879 on Build GRCh38
rs760455234 Chr.8: 41609894-41609894 on Build GRCh38
rs769715619 Chr.8: 41609844-41609844 on Build GRCh38
rs763844705 Chr.8: 41609895-41609895 on Build GRCh38
rs967952504 Chr.8: 41610013-41610013 on Build GRCh38
rs780693887 Chr.8: 41609914-41609914 on Build GRCh38
rs375193717 Chr.8: 41610001-41610001 on Build GRCh38
rs767043106 Chr.8: 41610006-41610006 on Build GRCh38
rs752825794 Chr.8: 41609984-41609984 on Build GRCh38
rs780497969 Chr.8: 41609818-41609818 on Build GRCh38
rs764815417 Chr.8: 41609997-41609997 on Build GRCh38

View Full Product Details