Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 12949506-12949779 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP44
Entrez Gene ID
9912
Gene Name
Rho GTPase activating protein 44
Gene Aliases
NPC-A-10, RICH2
UniGene
Hs.499758
Gene Chromosome Location
Chr.17: 12789512-12991644 on Build GRCh38
Transcripts
NM_001321164.1 NM_001321166.1 NM_001321167.1 NM_001321168.1 NM_014859.5 NR_135569.1 XM_005256891.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6603666 c.976G>A p.A326T Chr.17: 12949651-12949651 on Build GRCh38
5748716 c.974-2A>G p.? Chr.17: 12949647-12949647 on Build GRCh38
2737912 c.990C>T p.Y330Y Chr.17: 12949665-12949665 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs753057302 Chr.17: 12949666-12949666 on Build GRCh38
rs777549654 Chr.17: 12949715-12949715 on Build GRCh38
rs768521811 Chr.17: 12949606-12949606 on Build GRCh38
rs771920163 Chr.17: 12949600-12949600 on Build GRCh38
rs779838453 Chr.17: 12949710-12949710 on Build GRCh38
rs879015191 Chr.17: 12949591-12949591 on Build GRCh38
rs775221767 Chr.17: 12949601-12949601 on Build GRCh38
rs895722846 Chr.17: 12949542-12949542 on Build GRCh38
rs370611827 Chr.17: 12949622-12949622 on Build GRCh38
rs778494505 Chr.17: 12949670-12949670 on Build GRCh38
rs1033940381 Chr.17: 12949580-12949580 on Build GRCh38
rs965612808 Chr.17: 12949659-12949659 on Build GRCh38
rs376229335 Chr.17: 12949728-12949728 on Build GRCh38
rs557884352 Chr.17: 12949552-12949552 on Build GRCh38
rs772115775 Chr.17: 12949701-12949701 on Build GRCh38
rs776535986 Chr.17: 12949616-12949616 on Build GRCh38
rs778806555 Chr.17: 12949695-12949695 on Build GRCh38
rs747989058 Chr.17: 12949749-12949749 on Build GRCh38
rs370189247 Chr.17: 12949620-12949620 on Build GRCh38
rs887943524 Chr.17: 12949547-12949547 on Build GRCh38
rs1032347503 Chr.17: 12949682-12949682 on Build GRCh38
rs746129971 Chr.17: 12949700-12949700 on Build GRCh38
rs767237314 Chr.17: 12949740-12949740 on Build GRCh38
rs762473992 Chr.17: 12949623-12949623 on Build GRCh38
rs373346673 Chr.17: 12949677-12949677 on Build GRCh38
rs201487249 Chr.17: 12949669-12949669 on Build GRCh38
rs761738728 Chr.17: 12949738-12949738 on Build GRCh38
rs767998458 Chr.17: 12949665-12949665 on Build GRCh38
rs896712588 Chr.17: 12949584-12949584 on Build GRCh38
rs1049263553 Chr.17: 12949539-12949539 on Build GRCh38
rs757405142 Chr.17: 12949690-12949690 on Build GRCh38
rs776589110 Chr.17: 12949742-12949742 on Build GRCh38
rs760606581 Chr.17: 12949603-12949603 on Build GRCh38
rs1036678697 Chr.17: 12949541-12949541 on Build GRCh38
rs761451453 Chr.17: 12949618-12949618 on Build GRCh38
rs1044206175 Chr.17: 12949549-12949549 on Build GRCh38

View Full Product Details