Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 151035494-151035755 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCB8
Entrez Gene ID
11194
Gene Name
ATP binding cassette subfamily B member 8
Gene Aliases
EST328128, M-ABC1, MABC1
UniGene
Hs.647118
Gene Chromosome Location
Chr.7: 151028422-151047782 on Build GRCh38
Transcripts
NM_001282291.1 NM_001282292.1 NM_001282293.1 NM_007188.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1195752 c.779G>A p.C260Y Chr.7: 151035594-151035594 on Build GRCh38
5195762 c.1008C>T p.L336L Chr.7: 151035712-151035712 on Build GRCh38
6281817 c.910C>T p.R304C Chr.7: 151035725-151035725 on Build GRCh38
6109603 c.570G>C p.T190T Chr.7: 151035649-151035649 on Build GRCh38
7173506 c.993G>A p.L331L Chr.7: 151035697-151035697 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs989154240 Chr.7: 151035609-151035609 on Build GRCh38
rs956292798 Chr.7: 151035589-151035589 on Build GRCh38
rs753416228 Chr.7: 151035539-151035539 on Build GRCh38
rs777937374 Chr.7: 151035664-151035664 on Build GRCh38
rs888355948 Chr.7: 151035712-151035712 on Build GRCh38
rs918397787 Chr.7: 151035693-151035693 on Build GRCh38
rs772287635 Chr.7: 151035579-151035579 on Build GRCh38
rs372442090 Chr.7: 151035560-151035560 on Build GRCh38
rs766562502 Chr.7: 151035709-151035709 on Build GRCh38
rs117517576 Chr.7: 151035547-151035547 on Build GRCh38
rs140337671 Chr.7: 151035700-151035700 on Build GRCh38
rs377745915 Chr.7: 151035722-151035722 on Build GRCh38
rs748519673 Chr.7: 151035565-151035565 on Build GRCh38
rs17545756 Chr.7: 151035725-151035725 on Build GRCh38
rs749370842 Chr.7: 151035559-151035559 on Build GRCh38
rs927018386 Chr.7: 151035526-151035526 on Build GRCh38
rs374301133 Chr.7: 151035636-151035636 on Build GRCh38
rs369818772 Chr.7: 151035587-151035587 on Build GRCh38
rs754096279 Chr.7: 151035714-151035714 on Build GRCh38
rs143198607 Chr.7: 151035588-151035588 on Build GRCh38
rs758961534 Chr.7: 151035663-151035663 on Build GRCh38
rs759480881 Chr.7: 151035542-151035542 on Build GRCh38
rs748557450 Chr.7: 151035662-151035662 on Build GRCh38
rs1005537406 Chr.7: 151035522-151035522 on Build GRCh38
rs766650530 Chr.7: 151035648-151035648 on Build GRCh38
rs774009449 Chr.7: 151035702-151035702 on Build GRCh38
rs997574158 Chr.7: 151035732-151035732 on Build GRCh38
rs915035173 Chr.7: 151035639-151035639 on Build GRCh38
rs1140977 Chr.7: 151035596-151035596 on Build GRCh38
rs769387562 Chr.7: 151035694-151035694 on Build GRCh38
rs570437547 Chr.7: 151035737-151035737 on Build GRCh38
rs939514487 Chr.7: 151035687-151035687 on Build GRCh38
rs576776514 Chr.7: 151035637-151035637 on Build GRCh38
rs190274756 Chr.7: 151035642-151035642 on Build GRCh38
rs760410721 Chr.7: 151035535-151035535 on Build GRCh38
rs866565338 Chr.7: 151035713-151035713 on Build GRCh38
rs929910278 Chr.7: 151035695-151035695 on Build GRCh38
rs770694537 Chr.7: 151035670-151035670 on Build GRCh38
rs749938933 Chr.7: 151035549-151035549 on Build GRCh38
rs752529638 Chr.7: 151035545-151035545 on Build GRCh38
rs776277200 Chr.7: 151035613-151035613 on Build GRCh38
rs564580952 Chr.7: 151035684-151035684 on Build GRCh38
rs775161864 Chr.7: 151035696-151035696 on Build GRCh38
rs76884999 Chr.7: 151035562-151035562 on Build GRCh38
rs753747262 Chr.7: 151035595-151035595 on Build GRCh38
rs184975044 Chr.7: 151035540-151035540 on Build GRCh38
rs758830222 Chr.7: 151035726-151035726 on Build GRCh38
rs747474835 Chr.7: 151035665-151035665 on Build GRCh38
rs151194300 Chr.7: 151035594-151035594 on Build GRCh38
rs762741020 Chr.7: 151035697-151035697 on Build GRCh38
rs531746653 Chr.7: 151035649-151035649 on Build GRCh38
rs755769690 Chr.7: 151035626-151035632 on Build GRCh38
rs201138111 Chr.7: 151035671-151035671 on Build GRCh38
rs41282730 Chr.7: 151035543-151035543 on Build GRCh38
rs372401926 Chr.7: 151035536-151035536 on Build GRCh38
rs965147474 Chr.7: 151035582-151035582 on Build GRCh38
rs756381592 Chr.7: 151035550-151035550 on Build GRCh38
rs542562581 Chr.7: 151035641-151035641 on Build GRCh38
rs758177945 Chr.7: 151035546-151035546 on Build GRCh38
rs779358458 Chr.7: 151035654-151035654 on Build GRCh38
rs762978153 Chr.7: 151035638-151035638 on Build GRCh38

View Full Product Details