Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35044521-35044758 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf131
Entrez Gene ID
138724
Gene Name
chromosome 9 open reading frame 131
Gene Aliases
-
UniGene
Hs.148250
Gene Chromosome Location
Chr.9: 35041095-35045991 on Build GRCh38
Transcripts
NM_001040410.2 NM_001040411.2 NM_001040412.2 NM_001287391.1 NM_203299.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4474813 c.1943C>T p.S648F Chr.9: 35044572-35044572 on Build GRCh38
6616817 c.1968C>T p.H656H Chr.9: 35044597-35044597 on Build GRCh38
6550073 c.1862C>T p.P621L Chr.9: 35044635-35044635 on Build GRCh38
3926745 c.1758A>G p.E586E Chr.9: 35044531-35044531 on Build GRCh38
6616818 c.1824C>T p.H608H Chr.9: 35044597-35044597 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776658699 Chr.9: 35044666-35044666 on Build GRCh38
rs568535177 Chr.9: 35044684-35044684 on Build GRCh38
rs775421201 Chr.9: 35044657-35044657 on Build GRCh38
rs761998592 Chr.9: 35044583-35044583 on Build GRCh38
rs530262362 Chr.9: 35044594-35044594 on Build GRCh38
rs771768349 Chr.9: 35044648-35044648 on Build GRCh38
rs750743229 Chr.9: 35044590-35044590 on Build GRCh38
rs16932163 Chr.9: 35044585-35044585 on Build GRCh38
rs201120816 Chr.9: 35044671-35044671 on Build GRCh38
rs116919703 Chr.9: 35044558-35044558 on Build GRCh38
rs775303097 Chr.9: 35044574-35044574 on Build GRCh38
rs370501361 Chr.9: 35044734-35044734 on Build GRCh38
rs551840845 Chr.9: 35044662-35044662 on Build GRCh38
rs763314542 Chr.9: 35044694-35044694 on Build GRCh38
rs186147830 Chr.9: 35044709-35044709 on Build GRCh38
rs917546402 Chr.9: 35044689-35044689 on Build GRCh38
rs376129253 Chr.9: 35044728-35044728 on Build GRCh38
rs114611745 Chr.9: 35044629-35044629 on Build GRCh38
rs759289586 Chr.9: 35044571-35044571 on Build GRCh38
rs774264558 Chr.9: 35044564-35044564 on Build GRCh38
rs1021298531 Chr.9: 35044563-35044563 on Build GRCh38
rs761908957 Chr.9: 35044669-35044669 on Build GRCh38
rs746905841 Chr.9: 35044659-35044659 on Build GRCh38
rs766652649 Chr.9: 35044695-35044695 on Build GRCh38
rs776748517 Chr.9: 35044582-35044582 on Build GRCh38
rs777411018 Chr.9: 35044598-35044598 on Build GRCh38
rs772005930 Chr.9: 35044572-35044572 on Build GRCh38
rs991889285 Chr.9: 35044673-35044673 on Build GRCh38
rs146998208 Chr.9: 35044639-35044639 on Build GRCh38
rs768180306 Chr.9: 35044727-35044727 on Build GRCh38
rs180766598 Chr.9: 35044597-35044597 on Build GRCh38
rs748858328 Chr.9: 35044613-35044613 on Build GRCh38
rs375840215 Chr.9: 35044580-35044580 on Build GRCh38
rs764153564 Chr.9: 35044581-35044581 on Build GRCh38
rs752021973 Chr.9: 35044705-35044705 on Build GRCh38
rs115115421 Chr.9: 35044640-35044640 on Build GRCh38
rs756941093 Chr.9: 35044628-35044628 on Build GRCh38
rs777360972 Chr.9: 35044581-35044582 on Build GRCh38
rs766855776 Chr.9: 35044596-35044596 on Build GRCh38
rs765481416 Chr.9: 35044588-35044588 on Build GRCh38

View Full Product Details