Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 129301911-129302184 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HMCES
Entrez Gene ID
56941
Gene Name
5-hydroxymethylcytosine (hmC) binding, ES cell-specific
Gene Aliases
C3orf37, DC12, SRAPD1
UniGene
Hs.458320
Gene Chromosome Location
Chr.3: 129278806-129305293 on Build GRCh38
Transcripts
NM_001006109.1 NM_020187.2 XM_005247636.3 XM_005247637.3 XM_017006877.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1616866 c.641C>G p.P214R Chr.3: 129301955-129301955 on Build GRCh38
5863461 c.667G>A p.V223I Chr.3: 129301981-129301981 on Build GRCh38
4442484 c.776C>T p.S259L Chr.3: 129302090-129302090 on Build GRCh38
419846 c.788C>G p.T263S Chr.3: 129302102-129302102 on Build GRCh38
159605 c.749A>G p.H250R Chr.3: 129302063-129302063 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762467088 Chr.3: 129302004-129302004 on Build GRCh38
rs963798831 Chr.3: 129302064-129302064 on Build GRCh38
rs748269390 Chr.3: 129301975-129301975 on Build GRCh38
rs569388062 Chr.3: 129301940-129301940 on Build GRCh38
rs779281632 Chr.3: 129302087-129302087 on Build GRCh38
rs1010511616 Chr.3: 129302006-129302006 on Build GRCh38
rs761196025 Chr.3: 129302146-129302146 on Build GRCh38
rs753658471 Chr.3: 129301955-129301955 on Build GRCh38
rs6765930 Chr.3: 129301935-129301935 on Build GRCh38
rs903569658 Chr.3: 129302151-129302151 on Build GRCh38
rs749565505 Chr.3: 129301947-129301947 on Build GRCh38
rs770605530 Chr.3: 129302105-129302105 on Build GRCh38
rs754958338 Chr.3: 129302077-129302077 on Build GRCh38
rs776112604 Chr.3: 129301992-129301992 on Build GRCh38
rs200715890 Chr.3: 129302091-129302091 on Build GRCh38
rs752139104 Chr.3: 129301998-129301998 on Build GRCh38
rs780137547 Chr.3: 129301941-129301941 on Build GRCh38
rs773572230 Chr.3: 129302143-129302143 on Build GRCh38
rs758745235 Chr.3: 129301994-129301994 on Build GRCh38
rs372612340 Chr.3: 129302104-129302104 on Build GRCh38
rs370771693 Chr.3: 129301990-129301990 on Build GRCh38
rs937387488 Chr.3: 129301967-129301967 on Build GRCh38
rs970647269 Chr.3: 129302131-129302131 on Build GRCh38
rs996306005 Chr.3: 129302117-129302117 on Build GRCh38
rs373554679 Chr.3: 129302080-129302080 on Build GRCh38
rs756404710 Chr.3: 129302029-129302029 on Build GRCh38
rs144681599 Chr.3: 129302159-129302159 on Build GRCh38
rs148008790 Chr.3: 129302125-129302125 on Build GRCh38
rs772066045 Chr.3: 129301983-129301983 on Build GRCh38
rs114999931 Chr.3: 129302010-129302010 on Build GRCh38
rs879871926 Chr.3: 129301965-129301965 on Build GRCh38
rs768001734 Chr.3: 129302008-129302008 on Build GRCh38
rs367763626 Chr.3: 129302093-129302093 on Build GRCh38
rs200290205 Chr.3: 129302090-129302090 on Build GRCh38
rs115922185 Chr.3: 129302124-129302124 on Build GRCh38
rs780450714 Chr.3: 129302052-129302052 on Build GRCh38
rs759257249 Chr.3: 129302114-129302114 on Build GRCh38
rs773293805 Chr.3: 129301985-129301985 on Build GRCh38
rs754176802 Chr.3: 129302072-129302072 on Build GRCh38
rs1008327342 Chr.3: 129302056-129302056 on Build GRCh38
rs770822720 Chr.3: 129301991-129301991 on Build GRCh38
rs1017917426 Chr.3: 129302058-129302058 on Build GRCh38
rs763635796 Chr.3: 129302128-129302128 on Build GRCh38
rs779087269 Chr.3: 129301964-129301964 on Build GRCh38
rs769887542 Chr.3: 129302151-129302152 on Build GRCh38
rs768596245 Chr.3: 129301954-129301954 on Build GRCh38
rs776434066 Chr.3: 129302107-129302107 on Build GRCh38

View Full Product Details