Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 102457368-102457629 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALKBH4
Entrez Gene ID
54784
Gene Name
alkB homolog 4, lysine demethylase
Gene Aliases
ABH4
UniGene
Hs.658598
Gene Chromosome Location
Chr.7: 102456220-102464874 on Build GRCh38
Transcripts
NM_017621.3 XM_005250464.2 XM_005250465.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3876730 c.698C>T p.A233V Chr.7: 102457605-102457605 on Build GRCh38
6052911 c.726G>A p.L242L Chr.7: 102457577-102457577 on Build GRCh38
3762290 c.740C>T p.A247V Chr.7: 102457563-102457563 on Build GRCh38
6027443 c.784G>C p.E262Q Chr.7: 102457519-102457519 on Build GRCh38
3745436 c.681C>T p.C227C Chr.7: 102457622-102457622 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1054777480 Chr.7: 102457481-102457481 on Build GRCh38
rs147621685 Chr.7: 102457523-102457523 on Build GRCh38
rs755424082 Chr.7: 102457576-102457576 on Build GRCh38
rs771299364 Chr.7: 102457530-102457530 on Build GRCh38
rs368338131 Chr.7: 102457400-102457400 on Build GRCh38
rs201800893 Chr.7: 102457529-102457529 on Build GRCh38
rs781216004 Chr.7: 102457424-102457424 on Build GRCh38
rs150409598 Chr.7: 102457588-102457588 on Build GRCh38
rs555966568 Chr.7: 102457567-102457567 on Build GRCh38
rs145230697 Chr.7: 102457519-102457519 on Build GRCh38
rs750623687 Chr.7: 102457405-102457405 on Build GRCh38
rs372663274 Chr.7: 102457522-102457522 on Build GRCh38
rs755629098 Chr.7: 102457509-102457509 on Build GRCh38
rs371943941 Chr.7: 102457423-102457423 on Build GRCh38
rs754992172 Chr.7: 102457587-102457587 on Build GRCh38
rs371297660 Chr.7: 102457391-102457391 on Build GRCh38
rs759354371 Chr.7: 102457507-102457507 on Build GRCh38
rs573529822 Chr.7: 102457538-102457538 on Build GRCh38
rs766260358 Chr.7: 102457410-102457410 on Build GRCh38
rs780395093 Chr.7: 102457464-102457464 on Build GRCh38
rs767584826 Chr.7: 102457558-102457558 on Build GRCh38
rs777078094 Chr.7: 102457482-102457482 on Build GRCh38
rs141300734 Chr.7: 102457399-102457399 on Build GRCh38
rs778050169 Chr.7: 102457591-102457591 on Build GRCh38
rs769890779 Chr.7: 102457492-102457492 on Build GRCh38
rs774524165 Chr.7: 102457593-102457593 on Build GRCh38
rs748948872 Chr.7: 102457438-102457438 on Build GRCh38
rs766639597 Chr.7: 102457501-102457501 on Build GRCh38
rs771311388 Chr.7: 102457592-102457592 on Build GRCh38
rs778737084 Chr.7: 102457594-102457594 on Build GRCh38
rs147434463 Chr.7: 102457454-102457454 on Build GRCh38
rs139810871 Chr.7: 102457502-102457502 on Build GRCh38
rs199913773 Chr.7: 102457390-102457390 on Build GRCh38
rs763229650 Chr.7: 102457497-102457497 on Build GRCh38
rs773361580 Chr.7: 102457496-102457496 on Build GRCh38
rs755610154 Chr.7: 102457429-102457429 on Build GRCh38
rs756909709 Chr.7: 102457512-102457512 on Build GRCh38
rs781726622 Chr.7: 102457579-102457579 on Build GRCh38
rs112279537 Chr.7: 102457562-102457562 on Build GRCh38
rs777325120 Chr.7: 102457510-102457510 on Build GRCh38
rs777331990 Chr.7: 102457431-102457431 on Build GRCh38
rs745957475 Chr.7: 102457596-102457596 on Build GRCh38
rs149696343 Chr.7: 102457568-102457568 on Build GRCh38
rs778482855 Chr.7: 102457516-102457516 on Build GRCh38
rs769167643 Chr.7: 102457476-102457476 on Build GRCh38
rs560580334 Chr.7: 102457491-102457491 on Build GRCh38
rs774673696 Chr.7: 102457531-102457531 on Build GRCh38
rs41275227 Chr.7: 102457563-102457563 on Build GRCh38
rs539806636 Chr.7: 102457508-102457508 on Build GRCh38
rs761966095 Chr.7: 102457428-102457428 on Build GRCh38
rs372291952 Chr.7: 102457520-102457520 on Build GRCh38
rs763860189 Chr.7: 102457565-102457565 on Build GRCh38
rs771836161 Chr.7: 102457597-102457597 on Build GRCh38
rs749962932 Chr.7: 102457571-102457571 on Build GRCh38
rs751610772 Chr.7: 102457420-102457420 on Build GRCh38
rs758173603 Chr.7: 102457408-102457408 on Build GRCh38
rs747309640 Chr.7: 102457475-102457475 on Build GRCh38

View Full Product Details