Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 41965340-41965593 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLI3
Entrez Gene ID
2737
Gene Name
GLI family zinc finger 3
Gene Aliases
ACLS, GCPS, GLI3-190, GLI3FL, PAP-A, PAPA, PAPA1, PAPB, PHS, PPDIV
UniGene
Hs.21509
Gene Chromosome Location
Chr.7: 41960949-42237019 on Build GRCh38
Transcripts
NM_000168.5 XM_011515274.2 XM_017011997.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3229912 c.3630T>G p.A1210A Chr.7: 41965443-41965443 on Build GRCh38
3229906 c.3687G>C p.L1229F Chr.7: 41965386-41965386 on Build GRCh38
331556 c.3572T>C p.F1191S Chr.7: 41965501-41965501 on Build GRCh38
4687146 c.3595G>A p.V1199I Chr.7: 41965478-41965478 on Build GRCh38
5630188 c.3674G>A p.G1225D Chr.7: 41965399-41965399 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765172622 Chr.7: 41965365-41965365 on Build GRCh38
rs757037250 Chr.7: 41965375-41965375 on Build GRCh38
rs759795464 Chr.7: 41965460-41965460 on Build GRCh38
rs780924905 Chr.7: 41965378-41965378 on Build GRCh38
rs199638498 Chr.7: 41965425-41965425 on Build GRCh38
rs138618219 Chr.7: 41965368-41965368 on Build GRCh38
rs771236755 Chr.7: 41965453-41965453 on Build GRCh38
rs761887854 Chr.7: 41965559-41965559 on Build GRCh38
rs112234184 Chr.7: 41965427-41965427 on Build GRCh38
rs376413286 Chr.7: 41965487-41965487 on Build GRCh38
rs779661748 Chr.7: 41965448-41965448 on Build GRCh38
rs369440387 Chr.7: 41965451-41965451 on Build GRCh38
rs982488568 Chr.7: 41965539-41965539 on Build GRCh38
rs769375582 Chr.7: 41965380-41965380 on Build GRCh38
rs773687251 Chr.7: 41965404-41965404 on Build GRCh38
rs765946554 Chr.7: 41965488-41965488 on Build GRCh38
rs143551701 Chr.7: 41965533-41965533 on Build GRCh38
rs775705092 Chr.7: 41965464-41965464 on Build GRCh38
rs377548569 Chr.7: 41965572-41965572 on Build GRCh38
rs778367264 Chr.7: 41965512-41965512 on Build GRCh38
rs764195505 Chr.7: 41965493-41965495 on Build GRCh38
rs773211288 Chr.7: 41965563-41965563 on Build GRCh38
rs1052830686 Chr.7: 41965530-41965530 on Build GRCh38
rs746224726 Chr.7: 41965524-41965524 on Build GRCh38
rs374797309 Chr.7: 41965452-41965452 on Build GRCh38
rs143942705 Chr.7: 41965564-41965564 on Build GRCh38
rs890431070 Chr.7: 41965455-41965455 on Build GRCh38
rs751373757 Chr.7: 41965438-41965438 on Build GRCh38
rs768573875 Chr.7: 41965545-41965545 on Build GRCh38
rs764694374 Chr.7: 41965566-41965566 on Build GRCh38
rs759635709 Chr.7: 41965423-41965423 on Build GRCh38
rs1021772969 Chr.7: 41965367-41965367 on Build GRCh38
rs751296776 Chr.7: 41965370-41965370 on Build GRCh38
rs118149040 Chr.7: 41965409-41965409 on Build GRCh38
rs948874998 Chr.7: 41965417-41965417 on Build GRCh38
rs750219400 Chr.7: 41965486-41965486 on Build GRCh38
rs767546154 Chr.7: 41965560-41965560 on Build GRCh38
rs771150664 Chr.7: 41965421-41965421 on Build GRCh38
rs754984727 Chr.7: 41965361-41965361 on Build GRCh38
rs543636524 Chr.7: 41965462-41965462 on Build GRCh38
rs376682835 Chr.7: 41965447-41965447 on Build GRCh38
rs781010668 Chr.7: 41965446-41965446 on Build GRCh38
rs776777058 Chr.7: 41965422-41965422 on Build GRCh38
rs536152310 Chr.7: 41965393-41965393 on Build GRCh38
rs369483231 Chr.7: 41965553-41965553 on Build GRCh38
rs150179670 Chr.7: 41965366-41965366 on Build GRCh38
rs764106412 Chr.7: 41965476-41965476 on Build GRCh38
rs776871680 Chr.7: 41965456-41965456 on Build GRCh38
rs749408059 Chr.7: 41965534-41965534 on Build GRCh38
rs541487979 Chr.7: 41965516-41965516 on Build GRCh38
rs553376455 Chr.7: 41965381-41965381 on Build GRCh38
rs747475411 Chr.7: 41965515-41965515 on Build GRCh38
rs748737328 Chr.7: 41965449-41965449 on Build GRCh38
rs145069572 Chr.7: 41965475-41965475 on Build GRCh38
rs761728304 Chr.7: 41965484-41965484 on Build GRCh38
rs200382337 Chr.7: 41965386-41965386 on Build GRCh38
rs970269178 Chr.7: 41965383-41965383 on Build GRCh38
rs753384480 Chr.7: 41965565-41965565 on Build GRCh38
rs754391600 Chr.7: 41965505-41965505 on Build GRCh38
rs757059523 Chr.7: 41965442-41965442 on Build GRCh38
rs577288886 Chr.7: 41965504-41965504 on Build GRCh38
rs1043144793 Chr.7: 41965435-41965435 on Build GRCh38
rs765262375 Chr.7: 41965424-41965424 on Build GRCh38
rs772438444 Chr.7: 41965403-41965403 on Build GRCh38
rs781588154 Chr.7: 41965519-41965519 on Build GRCh38

View Full Product Details