Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 127446528-127446795 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0408
Entrez Gene ID
9729
Gene Name
KIAA0408
Gene Aliases
-
UniGene
Hs.319247
Gene Chromosome Location
Chr.6: 127438406-127459390 on Build GRCh38
Transcripts
NM_014702.4 NM_001012279.2

Gene Symbol
SOGA3
Entrez Gene ID
387104
Gene Name
SOGA family member 3
Gene Aliases
C6orf174, dJ403A15.3
UniGene
Hs.319247
Gene Chromosome Location
Chr.6: 127438406-127519355 on Build GRCh38
Transcripts
NM_014702.4 NM_001012279.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6514367 c.1631C>T p.P544L Chr.6: 127446688-127446688 on Build GRCh38
7295234 c.1762T>C p.W588R Chr.6: 127446557-127446557 on Build GRCh38
1072994 c.1773T>C p.C591C Chr.6: 127446546-127446546 on Build GRCh38
3947981 c.1680T>C p.Y560Y Chr.6: 127446639-127446639 on Build GRCh38
1072995 c.1648C>T p.R550C Chr.6: 127446671-127446671 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767969756 Chr.6: 127446667-127446667 on Build GRCh38
rs765203412 Chr.6: 127446764-127446764 on Build GRCh38
rs779822528 Chr.6: 127446718-127446718 on Build GRCh38
rs761243388 Chr.6: 127446636-127446636 on Build GRCh38
rs146403375 Chr.6: 127446763-127446763 on Build GRCh38
rs17054687 Chr.6: 127446759-127446759 on Build GRCh38
rs771790738 Chr.6: 127446598-127446598 on Build GRCh38
rs749443832 Chr.6: 127446556-127446556 on Build GRCh38
rs367604179 Chr.6: 127446576-127446576 on Build GRCh38
rs781112554 Chr.6: 127446725-127446725 on Build GRCh38
rs779012971 Chr.6: 127446571-127446571 on Build GRCh38
rs768525449 Chr.6: 127446719-127446719 on Build GRCh38
rs779151231 Chr.6: 127446700-127446700 on Build GRCh38
rs746921568 Chr.6: 127446614-127446614 on Build GRCh38
rs772770879 Chr.6: 127446656-127446656 on Build GRCh38
rs561359691 Chr.6: 127446651-127446651 on Build GRCh38
rs998982732 Chr.6: 127446747-127446747 on Build GRCh38
rs779875574 Chr.6: 127446610-127446610 on Build GRCh38
rs758690216 Chr.6: 127446712-127446712 on Build GRCh38
rs951095025 Chr.6: 127446563-127446563 on Build GRCh38
rs188314558 Chr.6: 127446720-127446720 on Build GRCh38
rs964026065 Chr.6: 127446658-127446658 on Build GRCh38
rs761129644 Chr.6: 127446758-127446758 on Build GRCh38
rs373508586 Chr.6: 127446631-127446631 on Build GRCh38
rs780270394 Chr.6: 127446771-127446771 on Build GRCh38
rs377310449 Chr.6: 127446687-127446687 on Build GRCh38
rs769423249 Chr.6: 127446641-127446641 on Build GRCh38
rs776107466 Chr.6: 127446621-127446621 on Build GRCh38
rs920170122 Chr.6: 127446683-127446683 on Build GRCh38
rs747655022 Chr.6: 127446727-127446727 on Build GRCh38
rs941153948 Chr.6: 127446705-127446705 on Build GRCh38
rs144655044 Chr.6: 127446675-127446675 on Build GRCh38
rs112232003 Chr.6: 127446688-127446688 on Build GRCh38
rs368021660 Chr.6: 127446615-127446615 on Build GRCh38
rs372590924 Chr.6: 127446766-127446766 on Build GRCh38
rs776324772 Chr.6: 127446594-127446597 on Build GRCh38
rs750592208 Chr.6: 127446701-127446701 on Build GRCh38
rs368538507 Chr.6: 127446752-127446752 on Build GRCh38
rs753151175 Chr.6: 127446669-127446669 on Build GRCh38
rs757484814 Chr.6: 127446564-127446564 on Build GRCh38
rs759895913 Chr.6: 127446666-127446666 on Build GRCh38
rs764752989 Chr.6: 127446679-127446679 on Build GRCh38
rs770515505 Chr.6: 127446588-127446588 on Build GRCh38
rs78892092 Chr.6: 127446739-127446739 on Build GRCh38
rs758537917 Chr.6: 127446768-127446768 on Build GRCh38
rs772827319 Chr.6: 127446728-127446728 on Build GRCh38
rs541088934 Chr.6: 127446748-127446748 on Build GRCh38
rs770772164 Chr.6: 127446744-127446744 on Build GRCh38
rs867700436 Chr.6: 127446586-127446586 on Build GRCh38
rs909629121 Chr.6: 127446637-127446637 on Build GRCh38

View Full Product Details