Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27333556-27333819 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTF3C2
Entrez Gene ID
2976
Gene Name
general transcription factor IIIC subunit 2
Gene Aliases
TFIIIC-BETA, TFIIIC110
UniGene
Hs.745168
Gene Chromosome Location
Chr.2: 27325849-27357034 on Build GRCh38
Transcripts
NM_001035521.2 NM_001318909.1 NM_001521.3 XM_005264272.4 XM_011532801.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1407425 c.1682C>A p.A561D Chr.2: 27333705-27333705 on Build GRCh38
6158123 c.1625A>T p.Q542L Chr.2: 27333762-27333762 on Build GRCh38
5967081 c.1698delG p.R566fs*6 Chr.2: 27333689-27333689 on Build GRCh38
5708176 c.1625A>G p.Q542R Chr.2: 27333762-27333762 on Build GRCh38
3981886 c.1606C>G p.Q536E Chr.2: 27333781-27333781 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764756046 Chr.2: 27333585-27333585 on Build GRCh38
rs985599935 Chr.2: 27333593-27333593 on Build GRCh38
rs781284698 Chr.2: 27333631-27333634 on Build GRCh38
rs563155629 Chr.2: 27333632-27333632 on Build GRCh38
rs901137880 Chr.2: 27333627-27333627 on Build GRCh38
rs751372634 Chr.2: 27333617-27333617 on Build GRCh38
rs78067336 Chr.2: 27333618-27333618 on Build GRCh38
rs190907956 Chr.2: 27333587-27333587 on Build GRCh38
rs867978987 Chr.2: 27333703-27333703 on Build GRCh38
rs369271760 Chr.2: 27333787-27333787 on Build GRCh38
rs776939264 Chr.2: 27333704-27333704 on Build GRCh38
rs201079868 Chr.2: 27333645-27333645 on Build GRCh38
rs761491613 Chr.2: 27333715-27333715 on Build GRCh38
rs764844460 Chr.2: 27333719-27333719 on Build GRCh38
rs765753079 Chr.2: 27333737-27333737 on Build GRCh38
rs772521770 Chr.2: 27333680-27333680 on Build GRCh38
rs145384574 Chr.2: 27333677-27333677 on Build GRCh38
rs570796218 Chr.2: 27333689-27333689 on Build GRCh38
rs769086264 Chr.2: 27333692-27333692 on Build GRCh38
rs372929888 Chr.2: 27333734-27333734 on Build GRCh38
rs762442496 Chr.2: 27333736-27333736 on Build GRCh38
rs532102436 Chr.2: 27333645-27333646 on Build GRCh38
rs546864555 Chr.2: 27333755-27333755 on Build GRCh38
rs150484118 Chr.2: 27333586-27333586 on Build GRCh38
rs780023801 Chr.2: 27333595-27333595 on Build GRCh38
rs771250146 Chr.2: 27333639-27333639 on Build GRCh38
rs375837098 Chr.2: 27333751-27333751 on Build GRCh38
rs566132276 Chr.2: 27333795-27333795 on Build GRCh38
rs755413830 Chr.2: 27333605-27333605 on Build GRCh38
rs752519823 Chr.2: 27333760-27333760 on Build GRCh38
rs929908159 Chr.2: 27333797-27333797 on Build GRCh38
rs779273684 Chr.2: 27333783-27333783 on Build GRCh38
rs750352576 Chr.2: 27333619-27333619 on Build GRCh38
rs780804841 Chr.2: 27333620-27333620 on Build GRCh38
rs757613008 Chr.2: 27333768-27333768 on Build GRCh38

View Full Product Details