Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 41474835-41475094 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NYX
Entrez Gene ID
60506
Gene Name
nyctalopin
Gene Aliases
CLRP, CSNB1, CSNB1A, CSNB4, NBM1
UniGene
Hs.302019
Gene Chromosome Location
Chr.X: 41447460-41476414 on Build GRCh38
Transcripts
NM_022567.2 XM_017029709.1

SNP Detail

SNP ID Chromosome Location
rs978067833 Chr.X: 41475026-41475026 on Build GRCh38
rs1027149135 Chr.X: 41474930-41474930 on Build GRCh38
rs891839220 Chr.X: 41475029-41475029 on Build GRCh38
rs750591219 Chr.X: 41474861-41474861 on Build GRCh38
rs1003093901 Chr.X: 41474860-41474860 on Build GRCh38
rs996119622 Chr.X: 41474929-41474929 on Build GRCh38
rs753101555 Chr.X: 41475047-41475048 on Build GRCh38
rs762960396 Chr.X: 41474910-41474910 on Build GRCh38
rs761926242 Chr.X: 41474960-41474960 on Build GRCh38
rs750048192 Chr.X: 41475002-41475002 on Build GRCh38
rs1057515883 Chr.X: 41474971-41474971 on Build GRCh38
rs140602960 Chr.X: 41474873-41474873 on Build GRCh38
rs370733549 Chr.X: 41475051-41475051 on Build GRCh38
rs906348045 Chr.X: 41474993-41474993 on Build GRCh38
rs187741852 Chr.X: 41475006-41475006 on Build GRCh38
rs754907887 Chr.X: 41474936-41474936 on Build GRCh38
rs923893223 Chr.X: 41475044-41475044 on Build GRCh38
rs769001640 Chr.X: 41474854-41474854 on Build GRCh38
rs954270376 Chr.X: 41474935-41474935 on Build GRCh38
rs755307939 Chr.X: 41475012-41475012 on Build GRCh38
rs367952714 Chr.X: 41474911-41474911 on Build GRCh38
rs767111588 Chr.X: 41474962-41474962 on Build GRCh38
rs531963566 Chr.X: 41475014-41475014 on Build GRCh38
rs1011570912 Chr.X: 41475039-41475039 on Build GRCh38
rs779803836 Chr.X: 41474919-41474919 on Build GRCh38
rs1057515882 Chr.X: 41474915-41474915 on Build GRCh38
rs1057515884 Chr.X: 41475056-41475056 on Build GRCh38
rs751397898 Chr.X: 41474920-41474920 on Build GRCh38

View Full Product Details