Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41610173-41610412 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT16
Entrez Gene ID
3868
Gene Name
keratin 16
Gene Aliases
CK16, FNEPPK, K16, K1CP, KRT16A, NEPPK, PC1
UniGene
Hs.655160
Gene Chromosome Location
Chr.17: 41609778-41612827 on Build GRCh38
Transcripts
NM_005557.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5664974 c.1226G>T p.R409L Chr.17: 41610385-41610385 on Build GRCh38
5663686 c.1324G>A p.E442K Chr.17: 41610193-41610193 on Build GRCh38
7176247 c.1270G>A p.E424K Chr.17: 41610341-41610341 on Build GRCh38
3517491 c.1200C>G p.Y400* Chr.17: 41610411-41610411 on Build GRCh38
5573999 c.1286_1287insT p.S430fs*>45 Chr.17: 41610230-41610231 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761807016 Chr.17: 41610342-41610342 on Build GRCh38
rs760901329 Chr.17: 41610204-41610204 on Build GRCh38
rs748198038 Chr.17: 41610283-41610283 on Build GRCh38
rs766782137 Chr.17: 41610213-41610213 on Build GRCh38
rs565842111 Chr.17: 41610287-41610287 on Build GRCh38
rs267607407 Chr.17: 41610341-41610341 on Build GRCh38
rs386797103 Chr.17: 41610252-41610253 on Build GRCh38
rs750221600 Chr.17: 41610347-41610347 on Build GRCh38
rs568310923 Chr.17: 41610295-41610295 on Build GRCh38
rs751535304 Chr.17: 41610256-41610256 on Build GRCh38
rs773801621 Chr.17: 41610196-41610196 on Build GRCh38
rs777901534 Chr.17: 41610358-41610358 on Build GRCh38
rs112929229 Chr.17: 41610237-41610237 on Build GRCh38
rs762324547 Chr.17: 41610312-41610312 on Build GRCh38
rs751249671 Chr.17: 41610319-41610319 on Build GRCh38
rs755238938 Chr.17: 41610271-41610271 on Build GRCh38
rs774567335 Chr.17: 41610193-41610193 on Build GRCh38
rs762673256 Chr.17: 41610388-41610388 on Build GRCh38
rs766195354 Chr.17: 41610311-41610338 on Build GRCh38
rs150453468 Chr.17: 41610300-41610300 on Build GRCh38
rs771443776 Chr.17: 41610369-41610369 on Build GRCh38
rs56259134 Chr.17: 41610359-41610359 on Build GRCh38
rs745725822 Chr.17: 41610270-41610270 on Build GRCh38
rs756004119 Chr.17: 41610350-41610350 on Build GRCh38
rs763676345 Chr.17: 41610323-41610323 on Build GRCh38
rs148365727 Chr.17: 41610386-41610386 on Build GRCh38
rs549128698 Chr.17: 41610289-41610289 on Build GRCh38
rs751388606 Chr.17: 41610341-41610341 on Build GRCh38
rs756941943 Chr.17: 41610265-41610265 on Build GRCh38
rs772428048 Chr.17: 41610195-41610195 on Build GRCh38
rs763894959 Chr.17: 41610254-41610254 on Build GRCh38
rs560220901 Chr.17: 41610253-41610253 on Build GRCh38
rs138288025 Chr.17: 41610355-41610355 on Build GRCh38
rs7406899 Chr.17: 41610252-41610252 on Build GRCh38
rs757745536 Chr.17: 41610381-41610381 on Build GRCh38
rs369631543 Chr.17: 41610233-41610233 on Build GRCh38
rs762797152 Chr.17: 41610282-41610283 on Build GRCh38
rs776202401 Chr.17: 41610223-41610223 on Build GRCh38
rs759188113 Chr.17: 41610231-41610231 on Build GRCh38
rs370949361 Chr.17: 41610284-41610284 on Build GRCh38
rs111470587 Chr.17: 41610238-41610238 on Build GRCh38
rs775068482 Chr.17: 41610387-41610387 on Build GRCh38
rs377221209 Chr.17: 41610257-41610257 on Build GRCh38
rs367591548 Chr.17: 41610194-41610194 on Build GRCh38
rs934666354 Chr.17: 41610326-41610326 on Build GRCh38
rs527635390 Chr.17: 41610279-41610279 on Build GRCh38
rs368945986 Chr.17: 41610385-41610385 on Build GRCh38
rs781137610 Chr.17: 41610266-41610266 on Build GRCh38
rs138335461 Chr.17: 41610356-41610356 on Build GRCh38
rs759938445 Chr.17: 41610291-41610291 on Build GRCh38

View Full Product Details