Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 38116036-38116271 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLA2G6
Entrez Gene ID
8398
Gene Name
phospholipase A2 group VI
Gene Aliases
CaI-PLA2, GVI, INAD1, IPLA2-VIA, NBIA2, NBIA2A, NBIA2B, PARK14, PLA2, PNPLA9, iPLA2, iPLA2beta
UniGene
Hs.170479
Gene Chromosome Location
Chr.22: 38111495-38192099 on Build GRCh38
Transcripts
NM_001004426.1 NM_001199562.1 NM_003560.2 XM_005261764.2 XM_005261765.1 XM_005261766.1 XM_006724332.3 XM_011530422.1 XM_011530423.1 XM_011530424.1 XM_011530425.1 XM_017028981.1 XM_017028982.1 XM_017028983.1 XM_017028984.1 XM_017028985.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1416226 c.1860G>A p.R620R Chr.22: 38116094-38116094 on Build GRCh38
3554344 c.1770C>T p.D590D Chr.22: 38116184-38116184 on Build GRCh38
4835329 c.1877C>T p.S626L Chr.22: 38116077-38116077 on Build GRCh38
4834061 c.1864C>G p.P622A Chr.22: 38116090-38116090 on Build GRCh38
726326 c.1749G>C p.M583I Chr.22: 38116205-38116205 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762533381 Chr.22: 38116128-38116128 on Build GRCh38
rs766703812 Chr.22: 38116233-38116233 on Build GRCh38
rs753504383 Chr.22: 38116138-38116138 on Build GRCh38
rs778516402 Chr.22: 38116148-38116148 on Build GRCh38
rs976845936 Chr.22: 38116112-38116112 on Build GRCh38
rs760792386 Chr.22: 38116087-38116087 on Build GRCh38
rs756996512 Chr.22: 38116147-38116147 on Build GRCh38
rs777465788 Chr.22: 38116136-38116136 on Build GRCh38
rs11570751 Chr.22: 38116237-38116237 on Build GRCh38
rs773374273 Chr.22: 38116223-38116223 on Build GRCh38
rs200117092 Chr.22: 38116119-38116119 on Build GRCh38
rs746984683 Chr.22: 38116175-38116175 on Build GRCh38
rs368008077 Chr.22: 38116156-38116156 on Build GRCh38
rs765536809 Chr.22: 38116114-38116114 on Build GRCh38
rs1043378899 Chr.22: 38116183-38116183 on Build GRCh38
rs370347639 Chr.22: 38116067-38116067 on Build GRCh38
rs753414506 Chr.22: 38116241-38116241 on Build GRCh38
rs867943649 Chr.22: 38116057-38116057 on Build GRCh38
rs776210772 Chr.22: 38116153-38116154 on Build GRCh38
rs756906527 Chr.22: 38116244-38116244 on Build GRCh38
rs768681156 Chr.22: 38116177-38116177 on Build GRCh38
rs763365476 Chr.22: 38116230-38116230 on Build GRCh38
rs767732261 Chr.22: 38116079-38116079 on Build GRCh38
rs776713955 Chr.22: 38116182-38116182 on Build GRCh38
rs149712244 Chr.22: 38116155-38116155 on Build GRCh38
rs762053663 Chr.22: 38116108-38116108 on Build GRCh38
rs753792105 Chr.22: 38116120-38116120 on Build GRCh38
rs146697329 Chr.22: 38116208-38116208 on Build GRCh38
rs139579057 Chr.22: 38116105-38116105 on Build GRCh38
rs745747130 Chr.22: 38116065-38116065 on Build GRCh38
rs764204393 Chr.22: 38116099-38116099 on Build GRCh38
rs587784338 Chr.22: 38116200-38116200 on Build GRCh38
rs775354024 Chr.22: 38116077-38116077 on Build GRCh38
rs924445048 Chr.22: 38116231-38116231 on Build GRCh38
rs200061943 Chr.22: 38116251-38116251 on Build GRCh38
rs750089741 Chr.22: 38116249-38116249 on Build GRCh38
rs761963849 Chr.22: 38116193-38116193 on Build GRCh38
rs199540781 Chr.22: 38116129-38116129 on Build GRCh38
rs763652863 Chr.22: 38116240-38116240 on Build GRCh38
rs760155875 Chr.22: 38116239-38116239 on Build GRCh38
rs780015355 Chr.22: 38116171-38116171 on Build GRCh38
rs34112430 Chr.22: 38116108-38116109 on Build GRCh38

View Full Product Details