Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 74129438-74129695 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO15
Entrez Gene ID
201456
Gene Name
F-box protein 15
Gene Aliases
FBX15
UniGene
Hs.664011
Gene Chromosome Location
Chr.18: 74073343-74147865 on Build GRCh38
Transcripts
NM_001142958.1 NM_152676.2 XM_011525856.1 XM_011525858.2 XM_011525859.1 XM_011525860.1 XM_017025603.1 XM_017025604.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6195549 c.405C>T p.I135I Chr.18: 74129557-74129557 on Build GRCh38
6149064 c.480A>G p.L160L Chr.18: 74129482-74129482 on Build GRCh38
6149062 c.449T>C p.V150A Chr.18: 74129513-74129513 on Build GRCh38
4073205 c.400T>C p.Y134H Chr.18: 74129562-74129562 on Build GRCh38
3891317 c.701C>T p.P234L Chr.18: 74129489-74129489 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770666245 Chr.18: 74129469-74129470 on Build GRCh38
rs750086635 Chr.18: 74129482-74129482 on Build GRCh38
rs142168799 Chr.18: 74129555-74129555 on Build GRCh38
rs375081838 Chr.18: 74129634-74129634 on Build GRCh38
rs747521191 Chr.18: 74129530-74129530 on Build GRCh38
rs201209798 Chr.18: 74129608-74129608 on Build GRCh38
rs201555346 Chr.18: 74129498-74129498 on Build GRCh38
rs751482531 Chr.18: 74129478-74129478 on Build GRCh38
rs764045183 Chr.18: 74129476-74129476 on Build GRCh38
rs199653489 Chr.18: 74129592-74129592 on Build GRCh38
rs113539425 Chr.18: 74129481-74129481 on Build GRCh38
rs753045811 Chr.18: 74129506-74129506 on Build GRCh38
rs200608267 Chr.18: 74129659-74129660 on Build GRCh38
rs977352310 Chr.18: 74129584-74129584 on Build GRCh38
rs769948142 Chr.18: 74129554-74129554 on Build GRCh38
rs753463240 Chr.18: 74129475-74129475 on Build GRCh38
rs755893773 Chr.18: 74129491-74129491 on Build GRCh38
rs764428805 Chr.18: 74129647-74129647 on Build GRCh38
rs757319963 Chr.18: 74129543-74129543 on Build GRCh38
rs775561918 Chr.18: 74129655-74129656 on Build GRCh38
rs1014279955 Chr.18: 74129510-74129510 on Build GRCh38
rs776325378 Chr.18: 74129623-74129624 on Build GRCh38
rs560347642 Chr.18: 74129620-74129620 on Build GRCh38
rs1022110284 Chr.18: 74129527-74129527 on Build GRCh38
rs758709331 Chr.18: 74129514-74129514 on Build GRCh38
rs745952070 Chr.18: 74129549-74129549 on Build GRCh38
rs138656692 Chr.18: 74129467-74129467 on Build GRCh38
rs111726894 Chr.18: 74129479-74129479 on Build GRCh38
rs370050834 Chr.18: 74129652-74129652 on Build GRCh38
rs778316161 Chr.18: 74129521-74129521 on Build GRCh38
rs992799237 Chr.18: 74129631-74129631 on Build GRCh38
rs781316620 Chr.18: 74129548-74129548 on Build GRCh38
rs199845878 Chr.18: 74129655-74129655 on Build GRCh38
rs76724179 Chr.18: 74129665-74129666 on Build GRCh38
rs374957246 Chr.18: 74129633-74129633 on Build GRCh38
rs536223754 Chr.18: 74129624-74129625 on Build GRCh38
rs749069347 Chr.18: 74129588-74129588 on Build GRCh38
rs565366800 Chr.18: 74129484-74129484 on Build GRCh38
rs75271717 Chr.18: 74129666-74129666 on Build GRCh38
rs761713454 Chr.18: 74129595-74129595 on Build GRCh38
rs1031715352 Chr.18: 74129598-74129598 on Build GRCh38
rs143154340 Chr.18: 74129556-74129556 on Build GRCh38
rs866325557 Chr.18: 74129490-74129490 on Build GRCh38
rs758770166 Chr.18: 74129641-74129641 on Build GRCh38
rs781674138 Chr.18: 74129659-74129659 on Build GRCh38
rs760272575 Chr.18: 74129627-74129627 on Build GRCh38
rs769602084 Chr.18: 74129625-74129625 on Build GRCh38
rs764282745 Chr.18: 74129654-74129655 on Build GRCh38
rs76675976 Chr.18: 74129667-74129667 on Build GRCh38
rs757696291 Chr.18: 74129653-74129653 on Build GRCh38

View Full Product Details