Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 113882635-113882908 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRAMD1C
Entrez Gene ID
54762
Gene Name
GRAM domain containing 1C
Gene Aliases
-
UniGene
Hs.24583
Gene Chromosome Location
Chr.3: 113828182-113947174 on Build GRCh38
Transcripts
NM_017577.4 XM_005247546.2 XM_005247547.1 XM_011512930.1 XM_017006646.1 XM_017006647.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6162634 c.487G>T p.D163Y Chr.3: 113882779-113882779 on Build GRCh38
4688378 c.535G>T p.D179Y Chr.3: 113882827-113882827 on Build GRCh38
1036568 c.527T>C p.V176A Chr.3: 113882819-113882819 on Build GRCh38
4688379 c.537T>C p.D179D Chr.3: 113882829-113882829 on Build GRCh38
3426902 c.524A>C p.N175T Chr.3: 113882816-113882816 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375932820 Chr.3: 113882716-113882716 on Build GRCh38
rs113884439 Chr.3: 113882772-113882772 on Build GRCh38
rs374765529 Chr.3: 113882775-113882775 on Build GRCh38
rs758017272 Chr.3: 113882787-113882800 on Build GRCh38
rs759022966 Chr.3: 113882808-113882808 on Build GRCh38
rs764168207 Chr.3: 113882833-113882833 on Build GRCh38
rs201025449 Chr.3: 113882758-113882758 on Build GRCh38
rs780267745 Chr.3: 113882843-113882843 on Build GRCh38
rs766013370 Chr.3: 113882804-113882804 on Build GRCh38
rs751030331 Chr.3: 113882706-113882706 on Build GRCh38
rs768633053 Chr.3: 113882774-113882774 on Build GRCh38
rs779732529 Chr.3: 113882829-113882833 on Build GRCh38
rs773542780 Chr.3: 113882794-113882794 on Build GRCh38
rs749341206 Chr.3: 113882763-113882763 on Build GRCh38
rs755258598 Chr.3: 113882873-113882873 on Build GRCh38
rs370197688 Chr.3: 113882745-113882745 on Build GRCh38
rs374091640 Chr.3: 113882828-113882828 on Build GRCh38
rs550805820 Chr.3: 113882719-113882719 on Build GRCh38
rs780170538 Chr.3: 113882748-113882748 on Build GRCh38
rs752317282 Chr.3: 113882822-113882822 on Build GRCh38
rs200996743 Chr.3: 113882778-113882778 on Build GRCh38
rs376858503 Chr.3: 113882863-113882863 on Build GRCh38
rs776167793 Chr.3: 113882806-113882806 on Build GRCh38
rs772625155 Chr.3: 113882779-113882779 on Build GRCh38
rs777702480 Chr.3: 113882711-113882711 on Build GRCh38
rs113821615 Chr.3: 113882677-113882677 on Build GRCh38
rs547997838 Chr.3: 113882717-113882717 on Build GRCh38
rs757025798 Chr.3: 113882837-113882837 on Build GRCh38
rs1007450847 Chr.3: 113882676-113882676 on Build GRCh38
rs754809610 Chr.3: 113882792-113882792 on Build GRCh38
rs751330821 Chr.3: 113882836-113882836 on Build GRCh38
rs760981656 Chr.3: 113882796-113882796 on Build GRCh38
rs904421726 Chr.3: 113882665-113882665 on Build GRCh38
rs756330431 Chr.3: 113882738-113882738 on Build GRCh38
rs140062638 Chr.3: 113882814-113882814 on Build GRCh38

View Full Product Details