Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 122422779-122423018 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSF2
Entrez Gene ID
3298
Gene Name
heat shock transcription factor 2
Gene Aliases
HSF 2, HSTF 2
UniGene
Hs.158195
Gene Chromosome Location
Chr.6: 122399551-122433119 on Build GRCh38
Transcripts
NM_001135564.1 NM_004506.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
128199 c.894C>T p.V298V Chr.6: 122422781-122422781 on Build GRCh38
3620070 c.1046A>T p.N349I Chr.6: 122422933-122422933 on Build GRCh38
6898781 c.1067G>A p.G356E Chr.6: 122422954-122422954 on Build GRCh38
4845647 c.900G>A p.Q300Q Chr.6: 122422787-122422787 on Build GRCh38
7225536 c.987A>T p.L329L Chr.6: 122422874-122422874 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369001271 Chr.6: 122422981-122422981 on Build GRCh38
rs986142448 Chr.6: 122422912-122422912 on Build GRCh38
rs779312725 Chr.6: 122422840-122422840 on Build GRCh38
rs776620124 Chr.6: 122422884-122422884 on Build GRCh38
rs746535519 Chr.6: 122422867-122422867 on Build GRCh38
rs45575931 Chr.6: 122422874-122422874 on Build GRCh38
rs764752770 Chr.6: 122422815-122422815 on Build GRCh38
rs762585761 Chr.6: 122422907-122422907 on Build GRCh38
rs757601713 Chr.6: 122422825-122422825 on Build GRCh38
rs769528436 Chr.6: 122422894-122422894 on Build GRCh38
rs984761289 Chr.6: 122422964-122422964 on Build GRCh38
rs754364398 Chr.6: 122422819-122422819 on Build GRCh38
rs775837251 Chr.6: 122422914-122422914 on Build GRCh38
rs1026876790 Chr.6: 122422928-122422928 on Build GRCh38
rs767861332 Chr.6: 122422910-122422910 on Build GRCh38
rs577591137 Chr.6: 122422833-122422833 on Build GRCh38
rs758102671 Chr.6: 122422854-122422854 on Build GRCh38
rs961260280 Chr.6: 122422876-122422876 on Build GRCh38
rs750807081 Chr.6: 122422979-122422979 on Build GRCh38
rs757804166 Chr.6: 122422955-122422955 on Build GRCh38
rs375274487 Chr.6: 122422975-122422975 on Build GRCh38
rs748090198 Chr.6: 122422888-122422888 on Build GRCh38
rs764237873 Chr.6: 122422937-122422937 on Build GRCh38
rs781236226 Chr.6: 122422904-122422904 on Build GRCh38
rs779617380 Chr.6: 122422859-122422859 on Build GRCh38
rs760903949 Chr.6: 122422924-122422924 on Build GRCh38
rs754350285 Chr.6: 122422950-122422950 on Build GRCh38
rs773087804 Chr.6: 122422903-122422903 on Build GRCh38
rs952591415 Chr.6: 122422963-122422963 on Build GRCh38
rs372249082 Chr.6: 122422846-122422846 on Build GRCh38

View Full Product Details