Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 226893958-226894217 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSEN2
Entrez Gene ID
5664
Gene Name
presenilin 2
Gene Aliases
AD3L, AD4, CMD1V, PS2, STM2
UniGene
Hs.25363
Gene Chromosome Location
Chr.1: 226870572-226903829 on Build GRCh38
Transcripts
NM_000447.2 NM_012486.2 XM_005273199.3 XM_017001835.1 XM_017001836.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6344290 c.1290+8G>T p.? Chr.1: 226894133-226894133 on Build GRCh38
4232746 c.1243A>T p.S415C Chr.1: 226894078-226894078 on Build GRCh38
5035543 c.1077C>T p.G359G Chr.1: 226894011-226894011 on Build GRCh38
3977116 c.1096G>C p.D366H Chr.1: 226894030-226894030 on Build GRCh38
6344289 c.1191+8G>T p.? Chr.1: 226894133-226894133 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770080595 Chr.1: 226894056-226894056 on Build GRCh38
rs201032114 Chr.1: 226893983-226893983 on Build GRCh38
rs145671982 Chr.1: 226894082-226894082 on Build GRCh38
rs766393518 Chr.1: 226894080-226894081 on Build GRCh38
rs753503617 Chr.1: 226894011-226894011 on Build GRCh38
rs377489312 Chr.1: 226894065-226894065 on Build GRCh38
rs751024219 Chr.1: 226894135-226894136 on Build GRCh38
rs563053551 Chr.1: 226894012-226894012 on Build GRCh38
rs771373304 Chr.1: 226894029-226894029 on Build GRCh38
rs201191318 Chr.1: 226893997-226893997 on Build GRCh38
rs199844009 Chr.1: 226894172-226894172 on Build GRCh38
rs575912514 Chr.1: 226894008-226894008 on Build GRCh38
rs199714082 Chr.1: 226894064-226894064 on Build GRCh38
rs750116511 Chr.1: 226894002-226894002 on Build GRCh38
rs781506963 Chr.1: 226894101-226894101 on Build GRCh38
rs781543554 Chr.1: 226894043-226894043 on Build GRCh38
rs200203079 Chr.1: 226894005-226894005 on Build GRCh38
rs143912759 Chr.1: 226894073-226894073 on Build GRCh38
rs2855562 Chr.1: 226894149-226894149 on Build GRCh38
rs1008797773 Chr.1: 226894174-226894174 on Build GRCh38
rs143549266 Chr.1: 226894097-226894097 on Build GRCh38
rs772255843 Chr.1: 226894140-226894140 on Build GRCh38
rs867544084 Chr.1: 226894094-226894094 on Build GRCh38
rs539696910 Chr.1: 226894062-226894062 on Build GRCh38
rs201772526 Chr.1: 226894083-226894083 on Build GRCh38
rs774513973 Chr.1: 226894066-226894066 on Build GRCh38
rs200034510 Chr.1: 226894144-226894144 on Build GRCh38
rs759194805 Chr.1: 226894157-226894157 on Build GRCh38
rs761844882 Chr.1: 226894068-226894068 on Build GRCh38
rs149172004 Chr.1: 226894090-226894090 on Build GRCh38
rs1042752261 Chr.1: 226894113-226894113 on Build GRCh38
rs753608837 Chr.1: 226894156-226894156 on Build GRCh38
rs779522932 Chr.1: 226894010-226894010 on Build GRCh38
rs369274454 Chr.1: 226894098-226894098 on Build GRCh38
rs760432548 Chr.1: 226894080-226894080 on Build GRCh38
rs780464244 Chr.1: 226894120-226894120 on Build GRCh38
rs201683508 Chr.1: 226894136-226894136 on Build GRCh38
rs551438819 Chr.1: 226894182-226894182 on Build GRCh38
rs142690225 Chr.1: 226894111-226894111 on Build GRCh38
rs201985133 Chr.1: 226893985-226893985 on Build GRCh38
rs763629376 Chr.1: 226894164-226894164 on Build GRCh38
rs750959403 Chr.1: 226894167-226894167 on Build GRCh38
rs774601959 Chr.1: 226894133-226894133 on Build GRCh38
rs202069038 Chr.1: 226894160-226894160 on Build GRCh38
rs377494557 Chr.1: 226894074-226894074 on Build GRCh38
rs768931743 Chr.1: 226894123-226894123 on Build GRCh38
rs115652716 Chr.1: 226894110-226894110 on Build GRCh38
rs746182253 Chr.1: 226894055-226894055 on Build GRCh38
rs1056738883 Chr.1: 226894081-226894081 on Build GRCh38
rs753499090 Chr.1: 226894087-226894087 on Build GRCh38
rs764832142 Chr.1: 226894159-226894159 on Build GRCh38
rs200852140 Chr.1: 226894175-226894175 on Build GRCh38
rs201093218 Chr.1: 226894026-226894026 on Build GRCh38
rs775674310 Chr.1: 226893982-226893982 on Build GRCh38
rs940775081 Chr.1: 226894007-226894007 on Build GRCh38
rs199550349 Chr.1: 226894086-226894086 on Build GRCh38
rs749533069 Chr.1: 226894122-226894122 on Build GRCh38
rs200432520 Chr.1: 226893989-226893989 on Build GRCh38
rs202197781 Chr.1: 226894143-226894143 on Build GRCh38
rs201891584 Chr.1: 226893998-226893998 on Build GRCh38

View Full Product Details