Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 106519795-106520028 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AIM1
Entrez Gene ID
202
Gene Name
absent in melanoma 1
Gene Aliases
CRYBG1, ST4
UniGene
Hs.724262
Gene Chromosome Location
Chr.6: 106360808-106570449 on Build GRCh38
Transcripts
NM_001624.3 XM_005266839.3 XM_017010333.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3154424 c.1395T>A p.S465S Chr.6: 106519827-106519827 on Build GRCh38
5095145 c.1551T>C p.L517L Chr.6: 106519983-106519983 on Build GRCh38
4899926 c.1429C>T p.P477S Chr.6: 106519861-106519861 on Build GRCh38
1187126 c.1447C>G p.Q483E Chr.6: 106519879-106519879 on Build GRCh38
450272 c.1562G>C p.G521A Chr.6: 106519994-106519994 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750082715 Chr.6: 106519931-106519931 on Build GRCh38
rs771931019 Chr.6: 106519854-106519854 on Build GRCh38
rs908518140 Chr.6: 106519835-106519835 on Build GRCh38
rs746060532 Chr.6: 106519977-106519977 on Build GRCh38
rs549196715 Chr.6: 106519936-106519936 on Build GRCh38
rs997002204 Chr.6: 106519851-106519851 on Build GRCh38
rs745517188 Chr.6: 106519852-106519852 on Build GRCh38
rs143832386 Chr.6: 106519859-106519859 on Build GRCh38
rs753101724 Chr.6: 106519905-106519905 on Build GRCh38
rs941622898 Chr.6: 106519844-106519844 on Build GRCh38
rs202072878 Chr.6: 106519895-106519895 on Build GRCh38
rs41305294 Chr.6: 106519872-106519872 on Build GRCh38
rs768677403 Chr.6: 106519860-106519860 on Build GRCh38
rs144881884 Chr.6: 106519956-106519956 on Build GRCh38
rs752790241 Chr.6: 106519865-106519865 on Build GRCh38
rs761067616 Chr.6: 106519906-106519906 on Build GRCh38
rs3747787 Chr.6: 106519903-106519903 on Build GRCh38
rs753269932 Chr.6: 106519971-106519972 on Build GRCh38
rs985547721 Chr.6: 106519848-106519848 on Build GRCh38
rs61734885 Chr.6: 106519958-106519958 on Build GRCh38
rs777119627 Chr.6: 106519841-106519841 on Build GRCh38
rs767865088 Chr.6: 106519904-106519904 on Build GRCh38
rs758911067 Chr.6: 106520005-106520006 on Build GRCh38
rs200154086 Chr.6: 106519881-106519881 on Build GRCh38
rs774500042 Chr.6: 106519901-106519901 on Build GRCh38
rs754338096 Chr.6: 106519918-106519918 on Build GRCh38
rs944027859 Chr.6: 106519879-106519879 on Build GRCh38
rs551716379 Chr.6: 106519968-106519968 on Build GRCh38
rs756723117 Chr.6: 106519921-106519921 on Build GRCh38
rs571526894 Chr.6: 106519976-106519976 on Build GRCh38
rs530129887 Chr.6: 106519916-106519916 on Build GRCh38
rs1051170511 Chr.6: 106519875-106519875 on Build GRCh38
rs780049885 Chr.6: 106519856-106519856 on Build GRCh38
rs959268225 Chr.6: 106519963-106519963 on Build GRCh38
rs778295701 Chr.6: 106519929-106519929 on Build GRCh38
rs756681428 Chr.6: 106519840-106519840 on Build GRCh38
rs778139113 Chr.6: 106519945-106519945 on Build GRCh38
rs148178366 Chr.6: 106519845-106519845 on Build GRCh38
rs144737726 Chr.6: 106519957-106519957 on Build GRCh38
rs185980551 Chr.6: 106520006-106520006 on Build GRCh38
rs545314721 Chr.6: 106519846-106519846 on Build GRCh38
rs764616273 Chr.6: 106519915-106519915 on Build GRCh38

View Full Product Details