Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161049545-161049770 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP30
Entrez Gene ID
257106
Gene Name
Rho GTPase activating protein 30
Gene Aliases
-
UniGene
Hs.389374
Gene Chromosome Location
Chr.1: 161046942-161069971 on Build GRCh38
Transcripts
NM_001025598.1 NM_001287600.1 NM_001287602.1 NM_181720.2 XM_005245070.2 XM_005245073.3 XM_011509391.2 XM_017000960.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
898439 c.1547C>T p.S516L Chr.1: 161049563-161049563 on Build GRCh38
5666276 c.1448C>T p.S483L Chr.1: 161049662-161049662 on Build GRCh38
4882731 c.1471C>A p.P491T Chr.1: 161049639-161049639 on Build GRCh38
5543725 c.1536C>T p.F512F Chr.1: 161049574-161049574 on Build GRCh38
898443 c.1503G>A p.S501S Chr.1: 161049607-161049607 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764200345 Chr.1: 161049604-161049604 on Build GRCh38
rs750713258 Chr.1: 161049687-161049687 on Build GRCh38
rs147106698 Chr.1: 161049633-161049633 on Build GRCh38
rs761754351 Chr.1: 161049607-161049607 on Build GRCh38
rs760987120 Chr.1: 161049679-161049679 on Build GRCh38
rs371644392 Chr.1: 161049657-161049657 on Build GRCh38
rs777485289 Chr.1: 161049659-161049659 on Build GRCh38
rs763243224 Chr.1: 161049709-161049709 on Build GRCh38
rs562996212 Chr.1: 161049682-161049682 on Build GRCh38
rs766313654 Chr.1: 161049719-161049719 on Build GRCh38
rs890753741 Chr.1: 161049693-161049693 on Build GRCh38
rs554009666 Chr.1: 161049631-161049631 on Build GRCh38
rs1039596140 Chr.1: 161049650-161049650 on Build GRCh38
rs370216018 Chr.1: 161049675-161049675 on Build GRCh38
rs909293664 Chr.1: 161049595-161049595 on Build GRCh38
rs151150229 Chr.1: 161049643-161049643 on Build GRCh38
rs772476087 Chr.1: 161049666-161049666 on Build GRCh38
rs781236066 Chr.1: 161049648-161049648 on Build GRCh38
rs548365507 Chr.1: 161049695-161049695 on Build GRCh38
rs758332758 Chr.1: 161049627-161049627 on Build GRCh38
rs145240665 Chr.1: 161049569-161049569 on Build GRCh38
rs535462719 Chr.1: 161049681-161049681 on Build GRCh38
rs922706357 Chr.1: 161049597-161049597 on Build GRCh38
rs374534204 Chr.1: 161049738-161049738 on Build GRCh38
rs754770296 Chr.1: 161049736-161049736 on Build GRCh38
rs540008446 Chr.1: 161049608-161049608 on Build GRCh38
rs978541512 Chr.1: 161049590-161049590 on Build GRCh38
rs778286823 Chr.1: 161049579-161049581 on Build GRCh38
rs370861606 Chr.1: 161049737-161049737 on Build GRCh38
rs933962888 Chr.1: 161049612-161049612 on Build GRCh38
rs751427963 Chr.1: 161049732-161049732 on Build GRCh38
rs144819608 Chr.1: 161049613-161049613 on Build GRCh38
rs753983424 Chr.1: 161049606-161049606 on Build GRCh38
rs748882098 Chr.1: 161049662-161049662 on Build GRCh38

View Full Product Details