Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42849035-42849282 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AOC2
Entrez Gene ID
314
Gene Name
amine oxidase, copper containing 2
Gene Aliases
DAO2, RAO
UniGene
Hs.143102
Gene Chromosome Location
Chr.17: 42844592-42850707 on Build GRCh38
Transcripts
NM_001158.3 NM_009590.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4066701 c.1638T>G p.A546A Chr.17: 42849135-42849135 on Build GRCh38
266011 c.1685G>A p.R562Q Chr.17: 42849182-42849182 on Build GRCh38
4066702 c.1742G>A p.R581H Chr.17: 42849239-42849239 on Build GRCh38
4926985 c.1642C>T p.P548S Chr.17: 42849139-42849139 on Build GRCh38
5852601 c.1733C>T p.P578L Chr.17: 42849230-42849230 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374132722 Chr.17: 42849099-42849099 on Build GRCh38
rs141488195 Chr.17: 42849261-42849261 on Build GRCh38
rs370118909 Chr.17: 42849079-42849079 on Build GRCh38
rs760695231 Chr.17: 42849163-42849163 on Build GRCh38
rs200995658 Chr.17: 42849062-42849062 on Build GRCh38
rs765645252 Chr.17: 42849181-42849181 on Build GRCh38
rs756757822 Chr.17: 42849220-42849220 on Build GRCh38
rs757943080 Chr.17: 42849085-42849085 on Build GRCh38
rs893058860 Chr.17: 42849214-42849214 on Build GRCh38
rs760025729 Chr.17: 42849246-42849246 on Build GRCh38
rs775503953 Chr.17: 42849229-42849229 on Build GRCh38
rs373057711 Chr.17: 42849060-42849060 on Build GRCh38
rs201776568 Chr.17: 42849162-42849162 on Build GRCh38
rs952170115 Chr.17: 42849101-42849101 on Build GRCh38
rs750530050 Chr.17: 42849078-42849078 on Build GRCh38
rs749112530 Chr.17: 42849230-42849230 on Build GRCh38
rs141210244 Chr.17: 42849227-42849227 on Build GRCh38
rs762194499 Chr.17: 42849151-42849151 on Build GRCh38
rs144886907 Chr.17: 42849105-42849105 on Build GRCh38
rs1002213011 Chr.17: 42849205-42849205 on Build GRCh38
rs765042636 Chr.17: 42849061-42849061 on Build GRCh38
rs565651756 Chr.17: 42849150-42849150 on Build GRCh38
rs768525345 Chr.17: 42849235-42849235 on Build GRCh38
rs758693648 Chr.17: 42849188-42849188 on Build GRCh38
rs776756237 Chr.17: 42849241-42849241 on Build GRCh38
rs755484979 Chr.17: 42849177-42849177 on Build GRCh38
rs767859340 Chr.17: 42849154-42849154 on Build GRCh38
rs745786817 Chr.17: 42849141-42849141 on Build GRCh38
rs766812474 Chr.17: 42849088-42849088 on Build GRCh38
rs767395208 Chr.17: 42849087-42849087 on Build GRCh38
rs199502196 Chr.17: 42849128-42849128 on Build GRCh38
rs975600373 Chr.17: 42849071-42849071 on Build GRCh38
rs767766603 Chr.17: 42849072-42849072 on Build GRCh38
rs777329590 Chr.17: 42849114-42849114 on Build GRCh38
rs779774526 Chr.17: 42849085-42849086 on Build GRCh38
rs138179430 Chr.17: 42849139-42849139 on Build GRCh38
rs758029863 Chr.17: 42849113-42849113 on Build GRCh38
rs776563901 Chr.17: 42849055-42849055 on Build GRCh38
rs200865307 Chr.17: 42849073-42849073 on Build GRCh38
rs780731878 Chr.17: 42849223-42849223 on Build GRCh38
rs562972964 Chr.17: 42849199-42849199 on Build GRCh38
rs143262935 Chr.17: 42849238-42849238 on Build GRCh38
rs544276929 Chr.17: 42849168-42849168 on Build GRCh38
rs746443779 Chr.17: 42849115-42849117 on Build GRCh38
rs745851722 Chr.17: 42849224-42849224 on Build GRCh38
rs148319810 Chr.17: 42849257-42849257 on Build GRCh38
rs962569474 Chr.17: 42849115-42849115 on Build GRCh38
rs755324690 Chr.17: 42849093-42849093 on Build GRCh38
rs551637666 Chr.17: 42849243-42849243 on Build GRCh38
rs746622808 Chr.17: 42849213-42849213 on Build GRCh38
rs949479414 Chr.17: 42849161-42849161 on Build GRCh38
rs746525146 Chr.17: 42849118-42849118 on Build GRCh38
rs762106450 Chr.17: 42849067-42849067 on Build GRCh38
rs760916686 Chr.17: 42849239-42849239 on Build GRCh38
rs754325554 Chr.17: 42849089-42849089 on Build GRCh38
rs200887039 Chr.17: 42849167-42849167 on Build GRCh38
rs769526425 Chr.17: 42849146-42849146 on Build GRCh38
rs928284722 Chr.17: 42849171-42849171 on Build GRCh38
rs753054416 Chr.17: 42849186-42849186 on Build GRCh38
rs1029119701 Chr.17: 42849100-42849100 on Build GRCh38
rs765735296 Chr.17: 42849254-42849254 on Build GRCh38

View Full Product Details