Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 110202361-110202632 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMMECR1
Entrez Gene ID
9949
Gene Name
Alport syndrome, mental retardation, midface hypoplasia and elliptocytosis chromosomal region gene 1
Gene Aliases
AMMERC1
UniGene
Hs.656243
Gene Chromosome Location
Chr.X: 110194186-110440233 on Build GRCh38
Transcripts
NM_001025580.1 NM_001171689.1 NM_015365.2

SNP Detail

SNP ID Chromosome Location
rs768604692 Chr.X: 110202536-110202536 on Build GRCh38
rs754001465 Chr.X: 110202435-110202437 on Build GRCh38
rs779348289 Chr.X: 110202420-110202420 on Build GRCh38
rs765510526 Chr.X: 110202564-110202565 on Build GRCh38
rs759367448 Chr.X: 110202436-110202436 on Build GRCh38
rs985011367 Chr.X: 110202576-110202576 on Build GRCh38
rs958661940 Chr.X: 110202462-110202462 on Build GRCh38
rs867942217 Chr.X: 110202517-110202517 on Build GRCh38
rs974317684 Chr.X: 110202484-110202484 on Build GRCh38
rs930165816 Chr.X: 110202507-110202507 on Build GRCh38
rs140856361 Chr.X: 110202486-110202486 on Build GRCh38
rs747190504 Chr.X: 110202471-110202471 on Build GRCh38
rs774399743 Chr.X: 110202564-110202564 on Build GRCh38
rs759627261 Chr.X: 110202547-110202548 on Build GRCh38
rs752938961 Chr.X: 110202586-110202590 on Build GRCh38
rs748689723 Chr.X: 110202421-110202421 on Build GRCh38
rs758447381 Chr.X: 110202424-110202424 on Build GRCh38
rs777963039 Chr.X: 110202465-110202465 on Build GRCh38
rs765240698 Chr.X: 110202577-110202577 on Build GRCh38
rs866602703 Chr.X: 110202466-110202466 on Build GRCh38
rs868514440 Chr.X: 110202467-110202467 on Build GRCh38
rs771116750 Chr.X: 110202477-110202477 on Build GRCh38
rs747363061 Chr.X: 110202515-110202515 on Build GRCh38

View Full Product Details