Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 151116924-151117157 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGAP3
Entrez Gene ID
116988
Gene Name
ArfGAP with GTPase domain, ankyrin repeat and PH domain 3
Gene Aliases
AGAP-3, CENTG3, CRAG, MRIP-1, cnt-g3
UniGene
Hs.647075
Gene Chromosome Location
Chr.7: 151085831-151144436 on Build GRCh38
Transcripts
NM_001042535.3 NM_001281300.1 NM_001308304.1 NM_001308305.1 NM_031946.6 XM_005249943.2 XM_011515780.1 XM_017011732.1 XM_017011733.1 XM_017011734.1 XM_017011735.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3431382 c.414C>T p.S138S Chr.7: 151117118-151117118 on Build GRCh38
7256844 c.422C>T p.S141L Chr.7: 151117126-151117126 on Build GRCh38
7256845 c.422C>T p.S141L Chr.7: 151117126-151117126 on Build GRCh38
3431383 c.414C>T p.S138S Chr.7: 151117118-151117118 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs540784828 Chr.7: 151117119-151117119 on Build GRCh38
rs377336159 Chr.7: 151117091-151117091 on Build GRCh38
rs943068061 Chr.7: 151116970-151116970 on Build GRCh38
rs367823906 Chr.7: 151117044-151117044 on Build GRCh38
rs774440561 Chr.7: 151117078-151117078 on Build GRCh38
rs1035021754 Chr.7: 151117068-151117068 on Build GRCh38
rs753135490 Chr.7: 151117051-151117052 on Build GRCh38
rs371496864 Chr.7: 151117046-151117046 on Build GRCh38
rs923047252 Chr.7: 151117007-151117007 on Build GRCh38
rs747882734 Chr.7: 151117079-151117079 on Build GRCh38
rs975771049 Chr.7: 151116994-151116994 on Build GRCh38
rs374060743 Chr.7: 151117090-151117090 on Build GRCh38
rs117148189 Chr.7: 151117047-151117047 on Build GRCh38
rs896446171 Chr.7: 151117081-151117081 on Build GRCh38
rs768768410 Chr.7: 151117074-151117074 on Build GRCh38
rs1002341658 Chr.7: 151116982-151116982 on Build GRCh38
rs910775942 Chr.7: 151116969-151116969 on Build GRCh38
rs370317322 Chr.7: 151117127-151117127 on Build GRCh38
rs749174435 Chr.7: 151117055-151117055 on Build GRCh38
rs369891624 Chr.7: 151117053-151117053 on Build GRCh38
rs1053241142 Chr.7: 151117020-151117020 on Build GRCh38
rs373601171 Chr.7: 151117130-151117130 on Build GRCh38
rs985027071 Chr.7: 151116965-151116965 on Build GRCh38
rs369444579 Chr.7: 151117048-151117048 on Build GRCh38
rs574862697 Chr.7: 151117052-151117058 on Build GRCh38
rs759850829 Chr.7: 151117112-151117112 on Build GRCh38
rs184989798 Chr.7: 151117038-151117038 on Build GRCh38
rs896143642 Chr.7: 151117011-151117011 on Build GRCh38
rs934434245 Chr.7: 151117010-151117010 on Build GRCh38
rs58651340 Chr.7: 151117051-151117051 on Build GRCh38
rs755453084 Chr.7: 151117051-151117051 on Build GRCh38
rs373183558 Chr.7: 151117050-151117050 on Build GRCh38

View Full Product Details