Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 14950485-14950758 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGDIB
Entrez Gene ID
397
Gene Name
Rho GDP dissociation inhibitor beta
Gene Aliases
D4, GDIA2, GDID4, LYGDI, Ly-GDI, RAP1GN1, RhoGDI2
UniGene
Hs.504877
Gene Chromosome Location
Chr.12: 14942015-14961770 on Build GRCh38
Transcripts
NM_001175.6 NM_001321420.1 NM_001321421.1 NM_001321422.1 NM_001321423.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
937585 c.153G>A p.T51T Chr.12: 14950560-14950560 on Build GRCh38
937586 c.147G>T p.K49N Chr.12: 14950566-14950566 on Build GRCh38
4040595 c.92C>T p.S31F Chr.12: 14950621-14950621 on Build GRCh38
6537618 c.61A>C p.K21Q Chr.12: 14950652-14950652 on Build GRCh38
2003167 c.23C>G p.P8R Chr.12: 14950690-14950690 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766315295 Chr.12: 14950687-14950687 on Build GRCh38
rs759475322 Chr.12: 14950505-14950505 on Build GRCh38
rs762075958 Chr.12: 14950613-14950613 on Build GRCh38
rs767575032 Chr.12: 14950524-14950524 on Build GRCh38
rs765540772 Chr.12: 14950639-14950639 on Build GRCh38
rs35503268 Chr.12: 14950726-14950727 on Build GRCh38
rs913863468 Chr.12: 14950708-14950708 on Build GRCh38
rs767661660 Chr.12: 14950620-14950620 on Build GRCh38
rs769639157 Chr.12: 14950508-14950508 on Build GRCh38
rs140719603 Chr.12: 14950641-14950641 on Build GRCh38
rs200119691 Chr.12: 14950560-14950560 on Build GRCh38
rs774246191 Chr.12: 14950669-14950669 on Build GRCh38
rs749179522 Chr.12: 14950660-14950660 on Build GRCh38
rs566164231 Chr.12: 14950586-14950586 on Build GRCh38
rs763215669 Chr.12: 14950703-14950703 on Build GRCh38
rs138036885 Chr.12: 14950513-14950513 on Build GRCh38
rs199554887 Chr.12: 14950509-14950509 on Build GRCh38
rs753254738 Chr.12: 14950692-14950692 on Build GRCh38
rs755804305 Chr.12: 14950728-14950728 on Build GRCh38
rs531618203 Chr.12: 14950592-14950592 on Build GRCh38
rs776717968 Chr.12: 14950688-14950688 on Build GRCh38
rs745592634 Chr.12: 14950656-14950656 on Build GRCh38
rs754388275 Chr.12: 14950556-14950556 on Build GRCh38
rs369959147 Chr.12: 14950585-14950585 on Build GRCh38
rs11541209 Chr.12: 14950626-14950626 on Build GRCh38
rs141671349 Chr.12: 14950581-14950581 on Build GRCh38
rs1028916280 Chr.12: 14950572-14950572 on Build GRCh38
rs1008601470 Chr.12: 14950582-14950582 on Build GRCh38
rs766761333 Chr.12: 14950548-14950548 on Build GRCh38
rs745341011 Chr.12: 14950584-14950584 on Build GRCh38
rs541241386 Chr.12: 14950541-14950541 on Build GRCh38
rs938725505 Chr.12: 14950507-14950507 on Build GRCh38
rs753815503 Chr.12: 14950605-14950605 on Build GRCh38
rs185689919 Chr.12: 14950510-14950510 on Build GRCh38
rs200666505 Chr.12: 14950557-14950557 on Build GRCh38
rs927585611 Chr.12: 14950715-14950715 on Build GRCh38
rs149654565 Chr.12: 14950670-14950670 on Build GRCh38
rs779661328 Chr.12: 14950733-14950733 on Build GRCh38
rs773817916 Chr.12: 14950561-14950561 on Build GRCh38
rs759999589 Chr.12: 14950689-14950689 on Build GRCh38
rs750489788 Chr.12: 14950534-14950534 on Build GRCh38
rs201399144 Chr.12: 14950655-14950655 on Build GRCh38
rs774299644 Chr.12: 14950611-14950611 on Build GRCh38
rs866356830 Chr.12: 14950690-14950690 on Build GRCh38
rs764587718 Chr.12: 14950705-14950705 on Build GRCh38
rs758839655 Chr.12: 14950645-14950645 on Build GRCh38
rs146450912 Chr.12: 14950637-14950637 on Build GRCh38
rs1042964582 Chr.12: 14950726-14950726 on Build GRCh38
rs755576194 Chr.12: 14950658-14950658 on Build GRCh38
rs190562188 Chr.12: 14950632-14950632 on Build GRCh38
rs773521470 Chr.12: 14950623-14950623 on Build GRCh38
rs780688334 Chr.12: 14950648-14950648 on Build GRCh38
rs762129203 Chr.12: 14950521-14950521 on Build GRCh38
rs375223965 Chr.12: 14950725-14950725 on Build GRCh38
rs200105682 Chr.12: 14950674-14950674 on Build GRCh38
rs779620521 Chr.12: 14950659-14950659 on Build GRCh38
rs754280234 Chr.12: 14950635-14950635 on Build GRCh38
rs756928111 Chr.12: 14950571-14950571 on Build GRCh38
rs2430711 Chr.12: 14950671-14950671 on Build GRCh38
rs768498974 Chr.12: 14950608-14950608 on Build GRCh38
rs899008445 Chr.12: 14950519-14950519 on Build GRCh38

View Full Product Details