Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 20640380-20640653 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSM1
Entrez Gene ID
116285
Gene Name
acyl-CoA synthetase medium-chain family member 1
Gene Aliases
BUCS1, MACS1
UniGene
Hs.306812
Gene Chromosome Location
Chr.16: 20623235-20697788 on Build GRCh38
Transcripts
NM_001318890.1 NM_052956.2 NR_134918.1 XM_006721016.2 XM_006721017.2 XM_006721018.2 XM_011545729.2 XM_011545730.2 XM_011545731.2 XM_011545732.2 XM_017022914.1 XM_017022915.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5717846 c.998G>A p.R333K Chr.16: 20640579-20640579 on Build GRCh38
968116 c.1039G>A p.V347M Chr.16: 20640538-20640538 on Build GRCh38
6025721 c.1030G>A p.G344R Chr.16: 20640547-20640547 on Build GRCh38
471431 c.1110G>A p.S370S Chr.16: 20640467-20640467 on Build GRCh38
702448 c.994A>G p.I332V Chr.16: 20640583-20640583 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777467315 Chr.16: 20640426-20640426 on Build GRCh38
rs764475898 Chr.16: 20640612-20640612 on Build GRCh38
rs761933838 Chr.16: 20640460-20640460 on Build GRCh38
rs977728394 Chr.16: 20640565-20640565 on Build GRCh38
rs574246403 Chr.16: 20640425-20640425 on Build GRCh38
rs372916427 Chr.16: 20640482-20640482 on Build GRCh38
rs745634207 Chr.16: 20640496-20640496 on Build GRCh38
rs756921981 Chr.16: 20640488-20640488 on Build GRCh38
rs772025673 Chr.16: 20640445-20640445 on Build GRCh38
rs1009227560 Chr.16: 20640625-20640625 on Build GRCh38
rs753200912 Chr.16: 20640478-20640478 on Build GRCh38
rs369221835 Chr.16: 20640501-20640501 on Build GRCh38
rs749354099 Chr.16: 20640568-20640568 on Build GRCh38
rs748758273 Chr.16: 20640544-20640544 on Build GRCh38
rs767833607 Chr.16: 20640519-20640519 on Build GRCh38
rs910009255 Chr.16: 20640626-20640626 on Build GRCh38
rs369054772 Chr.16: 20640451-20640451 on Build GRCh38
rs921366930 Chr.16: 20640594-20640594 on Build GRCh38
rs141054663 Chr.16: 20640500-20640500 on Build GRCh38
rs779717715 Chr.16: 20640547-20640547 on Build GRCh38
rs763091056 Chr.16: 20640466-20640466 on Build GRCh38
rs1050309800 Chr.16: 20640453-20640453 on Build GRCh38
rs778636434 Chr.16: 20640489-20640489 on Build GRCh38
rs774706434 Chr.16: 20640570-20640570 on Build GRCh38
rs761301535 Chr.16: 20640628-20640628 on Build GRCh38
rs753317504 Chr.16: 20640527-20640527 on Build GRCh38
rs377637288 Chr.16: 20640548-20640548 on Build GRCh38
rs545110583 Chr.16: 20640541-20640541 on Build GRCh38
rs893607442 Chr.16: 20640435-20640435 on Build GRCh38
rs940093049 Chr.16: 20640627-20640627 on Build GRCh38
rs748960028 Chr.16: 20640430-20640430 on Build GRCh38
rs761203831 Chr.16: 20640604-20640604 on Build GRCh38
rs774761233 Chr.16: 20640512-20640512 on Build GRCh38
rs150947761 Chr.16: 20640486-20640486 on Build GRCh38
rs777871947 Chr.16: 20640559-20640559 on Build GRCh38
rs745753186 Chr.16: 20640443-20640443 on Build GRCh38
rs768114625 Chr.16: 20640477-20640477 on Build GRCh38
rs777148297 Chr.16: 20640484-20640484 on Build GRCh38
rs565501213 Chr.16: 20640523-20640523 on Build GRCh38
rs759838963 Chr.16: 20640582-20640582 on Build GRCh38
rs1054905991 Chr.16: 20640578-20640578 on Build GRCh38
rs765523680 Chr.16: 20640498-20640498 on Build GRCh38
rs991636648 Chr.16: 20640538-20640538 on Build GRCh38
rs778517508 Chr.16: 20640539-20640539 on Build GRCh38
rs756841327 Chr.16: 20640565-20640565 on Build GRCh38
rs771904973 Chr.16: 20640497-20640497 on Build GRCh38
rs527828401 Chr.16: 20640555-20640555 on Build GRCh38
rs763221055 Chr.16: 20640504-20640504 on Build GRCh38
rs766792961 Chr.16: 20640467-20640467 on Build GRCh38
rs879223456 Chr.16: 20640543-20640543 on Build GRCh38
rs374754396 Chr.16: 20640442-20640442 on Build GRCh38
rs750135241 Chr.16: 20640540-20640540 on Build GRCh38
rs772423149 Chr.16: 20640597-20640597 on Build GRCh38
rs564768598 Chr.16: 20640505-20640505 on Build GRCh38
rs544937639 Chr.16: 20640440-20640440 on Build GRCh38
rs765295558 Chr.16: 20640461-20640461 on Build GRCh38
rs563014407 Chr.16: 20640462-20640462 on Build GRCh38
rs578181180 Chr.16: 20640468-20640468 on Build GRCh38
rs936021942 Chr.16: 20640571-20640571 on Build GRCh38
rs759963679 Chr.16: 20640518-20640518 on Build GRCh38
rs753176879 Chr.16: 20640520-20640520 on Build GRCh38
rs371287556 Chr.16: 20640601-20640601 on Build GRCh38
rs753469023 Chr.16: 20640485-20640485 on Build GRCh38
rs763610651 Chr.16: 20640526-20640526 on Build GRCh38
rs756798985 Chr.16: 20640533-20640533 on Build GRCh38
rs374267586 Chr.16: 20640503-20640503 on Build GRCh38
rs12444145 Chr.16: 20640616-20640616 on Build GRCh38

View Full Product Details