Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 109508699-109508950 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMIGO1
Entrez Gene ID
57463
Gene Name
adhesion molecule with Ig like domain 1
Gene Aliases
ALI2, AMIGO, AMIGO-1
UniGene
Hs.726479
Gene Chromosome Location
Chr.1: 109504178-109509742 on Build GRCh38
Transcripts
NM_020703.2 XM_011541812.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4661385 c.181A>G p.S61G Chr.1: 109508732-109508732 on Build GRCh38
4489592 c.34C>T p.L12F Chr.1: 109508879-109508879 on Build GRCh38
6599232 c.175T>C p.L59L Chr.1: 109508738-109508738 on Build GRCh38
1332461 c.121G>A p.A41T Chr.1: 109508792-109508792 on Build GRCh38
5024750 c.85C>T p.R29* Chr.1: 109508828-109508828 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775282824 Chr.1: 109508879-109508879 on Build GRCh38
rs767189663 Chr.1: 109508804-109508804 on Build GRCh38
rs763482556 Chr.1: 109508930-109508930 on Build GRCh38
rs756946213 Chr.1: 109508799-109508799 on Build GRCh38
rs139908524 Chr.1: 109508822-109508822 on Build GRCh38
rs745599368 Chr.1: 109508865-109508865 on Build GRCh38
rs140250244 Chr.1: 109508757-109508757 on Build GRCh38
rs200844103 Chr.1: 109508769-109508769 on Build GRCh38
rs758398955 Chr.1: 109508835-109508835 on Build GRCh38
rs909079720 Chr.1: 109508818-109508818 on Build GRCh38
rs771234042 Chr.1: 109508856-109508856 on Build GRCh38
rs41313278 Chr.1: 109508843-109508843 on Build GRCh38
rs776604823 Chr.1: 109508721-109508721 on Build GRCh38
rs751151577 Chr.1: 109508797-109508797 on Build GRCh38
rs779329833 Chr.1: 109508830-109508830 on Build GRCh38
rs760006404 Chr.1: 109508851-109508852 on Build GRCh38
rs985026706 Chr.1: 109508805-109508805 on Build GRCh38
rs894714260 Chr.1: 109508883-109508883 on Build GRCh38
rs147744049 Chr.1: 109508793-109508793 on Build GRCh38
rs146766120 Chr.1: 109508840-109508840 on Build GRCh38
rs747165516 Chr.1: 109508848-109508848 on Build GRCh38
rs145793775 Chr.1: 109508827-109508827 on Build GRCh38
rs200434051 Chr.1: 109508888-109508888 on Build GRCh38
rs780585630 Chr.1: 109508752-109508752 on Build GRCh38
rs148425119 Chr.1: 109508748-109508748 on Build GRCh38
rs769716146 Chr.1: 109508868-109508868 on Build GRCh38
rs759431964 Chr.1: 109508740-109508740 on Build GRCh38
rs754153484 Chr.1: 109508824-109508824 on Build GRCh38
rs375349763 Chr.1: 109508831-109508831 on Build GRCh38
rs17854056 Chr.1: 109508738-109508738 on Build GRCh38
rs542838804 Chr.1: 109508743-109508743 on Build GRCh38
rs374996680 Chr.1: 109508892-109508892 on Build GRCh38
rs777011950 Chr.1: 109508860-109508860 on Build GRCh38

View Full Product Details