Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 58610202-58610427 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM10
Entrez Gene ID
102
Gene Name
ADAM metallopeptidase domain 10
Gene Aliases
AD10, AD18, CD156c, CDw156, HsT18717, MADM, RAK, kuz
UniGene
Hs.172028
Gene Chromosome Location
Chr.15: 58595204-58749978 on Build GRCh38
Transcripts
NM_001110.3 NM_001320570.1 XM_017021861.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6001737 c.2014G>C p.E672Q Chr.15: 58610308-58610308 on Build GRCh38
1129166 c.1898A>G p.N633S Chr.15: 58610424-58610424 on Build GRCh38
700857 c.1931G>A p.R644Q Chr.15: 58610391-58610391 on Build GRCh38
2219432 c.1977delA p.A660fs*29 Chr.15: 58610345-58610345 on Build GRCh38
5653349 c.2019G>T p.W673C Chr.15: 58610303-58610303 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767527189 Chr.15: 58610276-58610276 on Build GRCh38
rs763144862 Chr.15: 58610269-58610269 on Build GRCh38
rs368571721 Chr.15: 58610267-58610267 on Build GRCh38
rs779647116 Chr.15: 58610391-58610391 on Build GRCh38
rs776564343 Chr.15: 58610268-58610268 on Build GRCh38
rs764498725 Chr.15: 58610324-58610324 on Build GRCh38
rs768703091 Chr.15: 58610394-58610394 on Build GRCh38
rs756605663 Chr.15: 58610322-58610322 on Build GRCh38
rs559729171 Chr.15: 58610239-58610239 on Build GRCh38
rs757941493 Chr.15: 58610390-58610390 on Build GRCh38
rs778661987 Chr.15: 58610359-58610359 on Build GRCh38
rs375834613 Chr.15: 58610392-58610392 on Build GRCh38
rs745466208 Chr.15: 58610378-58610378 on Build GRCh38
rs200845915 Chr.15: 58610282-58610282 on Build GRCh38
rs753205695 Chr.15: 58610288-58610288 on Build GRCh38
rs778332155 Chr.15: 58610237-58610237 on Build GRCh38
rs745349561 Chr.15: 58610238-58610238 on Build GRCh38
rs746887946 Chr.15: 58610265-58610265 on Build GRCh38
rs760720339 Chr.15: 58610281-58610282 on Build GRCh38
rs759683651 Chr.15: 58610275-58610275 on Build GRCh38
rs181056263 Chr.15: 58610330-58610330 on Build GRCh38
rs756896693 Chr.15: 58610332-58610332 on Build GRCh38
rs528456359 Chr.15: 58610272-58610272 on Build GRCh38
rs775595685 Chr.15: 58610259-58610259 on Build GRCh38

View Full Product Details