Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7688872-7689121 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WRAP53
Entrez Gene ID
55135
Gene Name
WD repeat containing antisense to TP53
Gene Aliases
DKCB3, TCAB1, WDR79
UniGene
Hs.408312
Gene Chromosome Location
Chr.17: 7686071-7703502 on Build GRCh38
Transcripts
NM_001143990.1 NM_001143991.1 NM_001143992.1 NM_018081.2 XM_011523952.2 XM_017024812.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5387436 c.322G>T p.E108* Chr.17: 7688970-7688970 on Build GRCh38
1480099 c.279C>T p.I93I Chr.17: 7688927-7688927 on Build GRCh38
6228675 c.235C>A p.L79M Chr.17: 7688883-7688883 on Build GRCh38
7096960 c.335G>A p.S112N Chr.17: 7688983-7688983 on Build GRCh38
6870854 c.227C>A p.S76Y Chr.17: 7688875-7688875 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769637437 Chr.17: 7688922-7688922 on Build GRCh38
rs17880740 Chr.17: 7689094-7689094 on Build GRCh38
rs767462997 Chr.17: 7688957-7688957 on Build GRCh38
rs751932334 Chr.17: 7688985-7688985 on Build GRCh38
rs770583524 Chr.17: 7689020-7689020 on Build GRCh38
rs759428765 Chr.17: 7688953-7688953 on Build GRCh38
rs749953879 Chr.17: 7689037-7689037 on Build GRCh38
rs755715290 Chr.17: 7689086-7689086 on Build GRCh38
rs747435394 Chr.17: 7688898-7688898 on Build GRCh38
rs750598622 Chr.17: 7688973-7688973 on Build GRCh38
rs745658108 Chr.17: 7689034-7689034 on Build GRCh38
rs751935581 Chr.17: 7689068-7689068 on Build GRCh38
rs774058987 Chr.17: 7689033-7689033 on Build GRCh38
rs141850830 Chr.17: 7688904-7688904 on Build GRCh38
rs777588420 Chr.17: 7689009-7689009 on Build GRCh38
rs774374559 Chr.17: 7688945-7688945 on Build GRCh38
rs909494338 Chr.17: 7688981-7688981 on Build GRCh38
rs759877669 Chr.17: 7689074-7689074 on Build GRCh38
rs554392556 Chr.17: 7688928-7688928 on Build GRCh38
rs767921886 Chr.17: 7689075-7689075 on Build GRCh38
rs929724373 Chr.17: 7689059-7689059 on Build GRCh38
rs755625682 Chr.17: 7689006-7689006 on Build GRCh38
rs148728256 Chr.17: 7688998-7688998 on Build GRCh38
rs749106865 Chr.17: 7688931-7688931 on Build GRCh38
rs144238223 Chr.17: 7688971-7688971 on Build GRCh38
rs771911772 Chr.17: 7689038-7689038 on Build GRCh38
rs34067256 Chr.17: 7689055-7689055 on Build GRCh38
rs766695794 Chr.17: 7688983-7688983 on Build GRCh38
rs770580910 Chr.17: 7688936-7688936 on Build GRCh38
rs763211473 Chr.17: 7689057-7689057 on Build GRCh38
rs766610610 Chr.17: 7689063-7689063 on Build GRCh38
rs1052321034 Chr.17: 7688926-7688927 on Build GRCh38
rs942160394 Chr.17: 7688986-7688986 on Build GRCh38
rs201340741 Chr.17: 7689043-7689043 on Build GRCh38
rs753373502 Chr.17: 7689100-7689100 on Build GRCh38
rs765624214 Chr.17: 7689071-7689072 on Build GRCh38
rs773992093 Chr.17: 7688997-7688997 on Build GRCh38
rs1043993272 Chr.17: 7689001-7689001 on Build GRCh38
rs904934289 Chr.17: 7689028-7689028 on Build GRCh38
rs753206220 Chr.17: 7689077-7689077 on Build GRCh38
rs780635910 Chr.17: 7688894-7688894 on Build GRCh38
rs150180394 Chr.17: 7688978-7688978 on Build GRCh38
rs765462600 Chr.17: 7688972-7688972 on Build GRCh38
rs775352941 Chr.17: 7689042-7689042 on Build GRCh38
rs757652943 Chr.17: 7689008-7689029 on Build GRCh38
rs372152816 Chr.17: 7688913-7688913 on Build GRCh38
rs776628490 Chr.17: 7688963-7688963 on Build GRCh38
rs781719919 Chr.17: 7689000-7689000 on Build GRCh38
rs780617002 Chr.17: 7688907-7688907 on Build GRCh38

View Full Product Details