Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 38395975-38396200 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH1B1
Entrez Gene ID
219
Gene Name
aldehyde dehydrogenase 1 family member B1
Gene Aliases
ALDH5, ALDHX
UniGene
Hs.436219
Gene Chromosome Location
Chr.9: 38392664-38398665 on Build GRCh38
Transcripts
NM_000692.4 XM_011517802.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
287233 c.242A>G p.K81R Chr.9: 38395990-38395990 on Build GRCh38
3930070 c.319C>T p.R107C Chr.9: 38396067-38396067 on Build GRCh38
5359515 c.281G>T p.R94L Chr.9: 38396029-38396029 on Build GRCh38
3930069 c.301C>T p.R101W Chr.9: 38396049-38396049 on Build GRCh38
6312175 c.309G>T p.R103R Chr.9: 38396057-38396057 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145597002 Chr.9: 38395998-38395998 on Build GRCh38
rs138929946 Chr.9: 38396154-38396154 on Build GRCh38
rs201556556 Chr.9: 38396095-38396095 on Build GRCh38
rs575584464 Chr.9: 38396055-38396055 on Build GRCh38
rs544611191 Chr.9: 38396176-38396176 on Build GRCh38
rs754908083 Chr.9: 38396050-38396050 on Build GRCh38
rs755851206 Chr.9: 38396125-38396125 on Build GRCh38
rs201981585 Chr.9: 38396047-38396047 on Build GRCh38
rs767408661 Chr.9: 38396114-38396114 on Build GRCh38
rs147069537 Chr.9: 38396024-38396024 on Build GRCh38
rs747533593 Chr.9: 38396166-38396166 on Build GRCh38
rs988866277 Chr.9: 38396175-38396175 on Build GRCh38
rs761172274 Chr.9: 38396025-38396025 on Build GRCh38
rs755939166 Chr.9: 38396056-38396056 on Build GRCh38
rs781160886 Chr.9: 38396053-38396053 on Build GRCh38
rs779060168 Chr.9: 38396136-38396136 on Build GRCh38
rs573017311 Chr.9: 38396076-38396076 on Build GRCh38
rs370716991 Chr.9: 38396129-38396129 on Build GRCh38
rs759092273 Chr.9: 38396111-38396111 on Build GRCh38
rs770857076 Chr.9: 38396067-38396067 on Build GRCh38
rs764953479 Chr.9: 38396028-38396028 on Build GRCh38
rs772311325 Chr.9: 38396153-38396153 on Build GRCh38
rs376279920 Chr.9: 38396032-38396032 on Build GRCh38
rs769088284 Chr.9: 38396089-38396089 on Build GRCh38
rs751229391 Chr.9: 38396035-38396035 on Build GRCh38
rs144035060 Chr.9: 38396031-38396031 on Build GRCh38
rs138473422 Chr.9: 38396029-38396029 on Build GRCh38
rs1033160085 Chr.9: 38396147-38396147 on Build GRCh38
rs766014764 Chr.9: 38396096-38396096 on Build GRCh38
rs199549604 Chr.9: 38396121-38396121 on Build GRCh38
rs752658129 Chr.9: 38396120-38396120 on Build GRCh38
rs772685418 Chr.9: 38396094-38396094 on Build GRCh38
rs749293011 Chr.9: 38396000-38396000 on Build GRCh38
rs201509595 Chr.9: 38396088-38396088 on Build GRCh38
rs751071218 Chr.9: 38396099-38396099 on Build GRCh38
rs745913809 Chr.9: 38396146-38396146 on Build GRCh38
rs774671053 Chr.9: 38396010-38396010 on Build GRCh38
rs200524448 Chr.9: 38396066-38396066 on Build GRCh38
rs151251560 Chr.9: 38396011-38396011 on Build GRCh38
rs540380490 Chr.9: 38396079-38396079 on Build GRCh38
rs772495848 Chr.9: 38396021-38396021 on Build GRCh38
rs148903407 Chr.9: 38395999-38395999 on Build GRCh38
rs2073478 Chr.9: 38396068-38396068 on Build GRCh38
rs777636836 Chr.9: 38396128-38396128 on Build GRCh38
rs2228093 Chr.9: 38396005-38396005 on Build GRCh38
rs141629640 Chr.9: 38396012-38396012 on Build GRCh38

View Full Product Details