Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 9211591-9211836 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD12
Entrez Gene ID
23253
Gene Name
ankyrin repeat domain 12
Gene Aliases
ANCO-2, ANCO1, GAC-1, Nbla00144
UniGene
Hs.464585
Gene Chromosome Location
Chr.18: 9136753-9285985 on Build GRCh38
Transcripts
NM_001083625.2 NM_001204056.1 NM_015208.4 XM_005258092.3 XM_005258093.3 XM_005258095.2 XM_005258096.2 XM_011525638.2 XM_011525639.2 XM_017025661.1 XM_017025662.1 XM_017025663.1 XM_017025664.1 XM_017025665.1 XM_017025666.1 XM_017025667.1 XM_017025668.1 XM_017025670.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4073648 c.546T>C p.N182N Chr.18: 9211678-9211678 on Build GRCh38
3527913 c.600A>G p.Q200Q Chr.18: 9211732-9211732 on Build GRCh38
5544477 c.470C>T p.P157L Chr.18: 9211602-9211602 on Build GRCh38
4661747 c.531A>C p.K177N Chr.18: 9211663-9211663 on Build GRCh38
3821795 c.607G>A p.E203K Chr.18: 9211739-9211739 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773508611 Chr.18: 9211682-9211682 on Build GRCh38
rs199693694 Chr.18: 9211669-9211669 on Build GRCh38
rs756701604 Chr.18: 9211756-9211756 on Build GRCh38
rs371683771 Chr.18: 9211753-9211753 on Build GRCh38
rs760955656 Chr.18: 9211694-9211694 on Build GRCh38
rs775039533 Chr.18: 9211645-9211645 on Build GRCh38
rs376494143 Chr.18: 9211752-9211752 on Build GRCh38
rs554670275 Chr.18: 9211802-9211802 on Build GRCh38
rs376367740 Chr.18: 9211801-9211801 on Build GRCh38
rs762702179 Chr.18: 9211712-9211712 on Build GRCh38
rs868166453 Chr.18: 9211791-9211791 on Build GRCh38
rs759133436 Chr.18: 9211660-9211661 on Build GRCh38
rs752520428 Chr.18: 9211703-9211703 on Build GRCh38
rs554017116 Chr.18: 9211758-9211758 on Build GRCh38
rs745384451 Chr.18: 9211650-9211650 on Build GRCh38
rs763985321 Chr.18: 9211741-9211741 on Build GRCh38
rs775921994 Chr.18: 9211625-9211625 on Build GRCh38
rs988620952 Chr.18: 9211616-9211616 on Build GRCh38
rs749674116 Chr.18: 9211627-9211627 on Build GRCh38
rs149795735 Chr.18: 9211634-9211634 on Build GRCh38
rs761946926 Chr.18: 9211653-9211653 on Build GRCh38
rs766577112 Chr.18: 9211702-9211702 on Build GRCh38
rs371533373 Chr.18: 9211615-9211615 on Build GRCh38

View Full Product Details