Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 4627256-4627493 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKAP3
Entrez Gene ID
10566
Gene Name
A-kinase anchoring protein 3
Gene Aliases
AKAP 110, AKAP110, CT82, FSP95, HEL159, PRKA3, SOB1
UniGene
Hs.98397
Gene Chromosome Location
Chr.12: 4615508-4649047 on Build GRCh38
Transcripts
NM_001278309.1 NM_006422.3 XM_005253664.2 XM_011520908.1 XM_011520909.1 XM_011520910.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2068275 c.1540G>C p.E514Q Chr.12: 4627362-4627362 on Build GRCh38
6243638 c.1437T>C p.F479F Chr.12: 4627465-4627465 on Build GRCh38
333542 c.1479C>G p.S493S Chr.12: 4627423-4627423 on Build GRCh38
1147015 c.1617G>T p.L539L Chr.12: 4627285-4627285 on Build GRCh38
3461076 c.1591G>A p.A531T Chr.12: 4627311-4627311 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs544521521 Chr.12: 4627446-4627446 on Build GRCh38
rs1043118003 Chr.12: 4627280-4627280 on Build GRCh38
rs767438859 Chr.12: 4627464-4627464 on Build GRCh38
rs767180671 Chr.12: 4627292-4627292 on Build GRCh38
rs150192660 Chr.12: 4627347-4627347 on Build GRCh38
rs1015988743 Chr.12: 4627462-4627462 on Build GRCh38
rs768381288 Chr.12: 4627439-4627439 on Build GRCh38
rs12366671 Chr.12: 4627403-4627403 on Build GRCh38
rs775065437 Chr.12: 4627294-4627294 on Build GRCh38
rs376352458 Chr.12: 4627351-4627351 on Build GRCh38
rs374081684 Chr.12: 4627448-4627448 on Build GRCh38
rs530706846 Chr.12: 4627414-4627414 on Build GRCh38
rs756373516 Chr.12: 4627353-4627353 on Build GRCh38
rs777662047 Chr.12: 4627359-4627359 on Build GRCh38
rs753366163 Chr.12: 4627278-4627279 on Build GRCh38
rs766908072 Chr.12: 4627337-4627337 on Build GRCh38
rs574267321 Chr.12: 4627404-4627404 on Build GRCh38
rs147621340 Chr.12: 4627381-4627381 on Build GRCh38
rs770962130 Chr.12: 4627371-4627371 on Build GRCh38
rs761261401 Chr.12: 4627323-4627323 on Build GRCh38
rs375126009 Chr.12: 4627408-4627408 on Build GRCh38
rs749128877 Chr.12: 4627360-4627360 on Build GRCh38
rs759161070 Chr.12: 4627288-4627288 on Build GRCh38
rs11063265 Chr.12: 4627465-4627465 on Build GRCh38
rs761716076 Chr.12: 4627280-4627281 on Build GRCh38
rs760193123 Chr.12: 4627419-4627419 on Build GRCh38
rs1006010014 Chr.12: 4627459-4627459 on Build GRCh38
rs755068981 Chr.12: 4627349-4627349 on Build GRCh38
rs1990312 Chr.12: 4627329-4627329 on Build GRCh38
rs773532348 Chr.12: 4627444-4627444 on Build GRCh38
rs765077715 Chr.12: 4627282-4627283 on Build GRCh38
rs765458308 Chr.12: 4627300-4627300 on Build GRCh38
rs750804755 Chr.12: 4627317-4627317 on Build GRCh38
rs370794366 Chr.12: 4627445-4627445 on Build GRCh38
rs762237719 Chr.12: 4627298-4627298 on Build GRCh38
rs375695630 Chr.12: 4627340-4627340 on Build GRCh38
rs748250818 Chr.12: 4627352-4627352 on Build GRCh38
rs760092151 Chr.12: 4627455-4627455 on Build GRCh38

View Full Product Details