Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 132896271-132896504 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSC1
Entrez Gene ID
7248
Gene Name
tuberous sclerosis 1
Gene Aliases
LAM, TSC
UniGene
Hs.370854
Gene Chromosome Location
Chr.9: 132891348-132945269 on Build GRCh38
Transcripts
NM_000368.4 NM_001162426.1 NM_001162427.1 XM_005272211.1 XM_006717271.1 XM_011518979.2 XM_017015096.1 XM_017015097.1 XM_017015098.1 XM_017015099.1 XM_017015100.1 XM_017015101.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6810207 c.3420G>A p.P1140P Chr.9: 132896310-132896310 on Build GRCh38
5063723 c.3237C>T p.G1079G Chr.9: 132896493-132896493 on Build GRCh38
6983157 c.3316G>A p.E1106K Chr.9: 132896414-132896414 on Build GRCh38
5546885 c.3302delA p.E1101fs*9 Chr.9: 132896428-132896428 on Build GRCh38
6961298 c.3296A>G p.K1099R Chr.9: 132896434-132896434 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751126355 Chr.9: 132896311-132896311 on Build GRCh38
rs118203751 Chr.9: 132896427-132896427 on Build GRCh38
rs745475737 Chr.9: 132896316-132896316 on Build GRCh38
rs775420987 Chr.9: 132896374-132896374 on Build GRCh38
rs773586317 Chr.9: 132896349-132896349 on Build GRCh38
rs767439431 Chr.9: 132896485-132896485 on Build GRCh38
rs779599439 Chr.9: 132896441-132896441 on Build GRCh38
rs878853967 Chr.9: 132896361-132896362 on Build GRCh38
rs756449737 Chr.9: 132896317-132896317 on Build GRCh38
rs751467429 Chr.9: 132896385-132896385 on Build GRCh38
rs759431801 Chr.9: 132896301-132896301 on Build GRCh38
rs952996030 Chr.9: 132896401-132896401 on Build GRCh38
rs140352085 Chr.9: 132896295-132896295 on Build GRCh38
rs118203750 Chr.9: 132896440-132896440 on Build GRCh38
rs774286125 Chr.9: 132896473-132896473 on Build GRCh38
rs796053467 Chr.9: 132896419-132896419 on Build GRCh38
rs550526986 Chr.9: 132896452-132896452 on Build GRCh38
rs200200869 Chr.9: 132896343-132896343 on Build GRCh38
rs796053463 Chr.9: 132896480-132896480 on Build GRCh38
rs754282309 Chr.9: 132896430-132896430 on Build GRCh38
rs762641110 Chr.9: 132896380-132896380 on Build GRCh38
rs764018144 Chr.9: 132896314-132896314 on Build GRCh38
rs760918561 Chr.9: 132896366-132896366 on Build GRCh38
rs773541137 Chr.9: 132896297-132896297 on Build GRCh38
rs769478982 Chr.9: 132896369-132896369 on Build GRCh38
rs886063621 Chr.9: 132896478-132896478 on Build GRCh38
rs764140399 Chr.9: 132896383-132896383 on Build GRCh38
rs568004490 Chr.9: 132896325-132896325 on Build GRCh38
rs774980129 Chr.9: 132896334-132896334 on Build GRCh38
rs761577419 Chr.9: 132896339-132896339 on Build GRCh38
rs761334590 Chr.9: 132896394-132896394 on Build GRCh38
rs769266225 Chr.9: 132896328-132896328 on Build GRCh38
rs758286878 Chr.9: 132896304-132896304 on Build GRCh38
rs35593170 Chr.9: 132896406-132896406 on Build GRCh38
rs763931959 Chr.9: 132896310-132896310 on Build GRCh38
rs767904247 Chr.9: 132896296-132896296 on Build GRCh38
rs776694051 Chr.9: 132896462-132896462 on Build GRCh38
rs116747861 Chr.9: 132896448-132896448 on Build GRCh38
rs574823234 Chr.9: 132896303-132896303 on Build GRCh38
rs772288584 Chr.9: 132896451-132896451 on Build GRCh38
rs751398082 Chr.9: 132896322-132896322 on Build GRCh38
rs876660132 Chr.9: 132896434-132896434 on Build GRCh38
rs772233665 Chr.9: 132896344-132896344 on Build GRCh38
rs771241484 Chr.9: 132896460-132896460 on Build GRCh38
rs118203752 Chr.9: 132896409-132896409 on Build GRCh38
rs201867031 Chr.9: 132896302-132896302 on Build GRCh38
rs118203753 Chr.9: 132896408-132896408 on Build GRCh38
rs756737864 Chr.9: 132896313-132896313 on Build GRCh38
rs747053008 Chr.9: 132896456-132896456 on Build GRCh38
rs761673589 Chr.9: 132896481-132896481 on Build GRCh38
rs762845573 Chr.9: 132896464-132896464 on Build GRCh38
rs749612772 Chr.9: 132896327-132896327 on Build GRCh38

View Full Product Details