Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 113062240-113062513 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL36RN
Entrez Gene ID
26525
Gene Name
interleukin 36 receptor antagonist
Gene Aliases
FIL1, FIL1(DELTA), FIL1D, IL-36Ra, IL1F5, IL1HY1, IL1L1, IL1RP3, IL36RA, PSORP, PSORS14
UniGene
Hs.516301
Gene Chromosome Location
Chr.2: 113058638-113064744 on Build GRCh38
Transcripts
NM_012275.2 NM_173170.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3565462 c.290G>A p.S97N Chr.2: 113062499-113062499 on Build GRCh38
3565460 c.258G>T p.M86I Chr.2: 113062467-113062467 on Build GRCh38
5532695 c.262C>T p.L88F Chr.2: 113062471-113062471 on Build GRCh38
3565461 c.284C>T p.S95F Chr.2: 113062493-113062493 on Build GRCh38
4133012 c.275C>A p.A92D Chr.2: 113062484-113062484 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs956637670 Chr.2: 113062394-113062394 on Build GRCh38
rs546539293 Chr.2: 113062356-113062356 on Build GRCh38
rs1029917786 Chr.2: 113062310-113062310 on Build GRCh38
rs977959390 Chr.2: 113062379-113062379 on Build GRCh38
rs769953299 Chr.2: 113062402-113062402 on Build GRCh38
rs996872677 Chr.2: 113062305-113062305 on Build GRCh38
rs964025748 Chr.2: 113062288-113062288 on Build GRCh38
rs200081179 Chr.2: 113062424-113062424 on Build GRCh38
rs201857918 Chr.2: 113062427-113062427 on Build GRCh38
rs537559199 Chr.2: 113062467-113062467 on Build GRCh38
rs973672613 Chr.2: 113062384-113062384 on Build GRCh38
rs778758453 Chr.2: 113062408-113062409 on Build GRCh38
rs749290931 Chr.2: 113062314-113062320 on Build GRCh38
rs886054767 Chr.2: 113062457-113062457 on Build GRCh38
rs775797643 Chr.2: 113062425-113062425 on Build GRCh38
rs144182857 Chr.2: 113062454-113062454 on Build GRCh38
rs571589576 Chr.2: 113062273-113062273 on Build GRCh38
rs771319833 Chr.2: 113062414-113062414 on Build GRCh38
rs369259981 Chr.2: 113062471-113062471 on Build GRCh38
rs746109701 Chr.2: 113062486-113062486 on Build GRCh38
rs772287603 Chr.2: 113062489-113062489 on Build GRCh38
rs748120723 Chr.2: 113062303-113062303 on Build GRCh38
rs778024459 Chr.2: 113062409-113062409 on Build GRCh38
rs1042989548 Chr.2: 113062455-113062455 on Build GRCh38
rs938489566 Chr.2: 113062432-113062432 on Build GRCh38
rs778949379 Chr.2: 113062474-113062474 on Build GRCh38
rs977901589 Chr.2: 113062263-113062263 on Build GRCh38
rs998871655 Chr.2: 113062487-113062487 on Build GRCh38
rs774774549 Chr.2: 113062422-113062422 on Build GRCh38
rs751374265 Chr.2: 113062282-113062282 on Build GRCh38
rs756170880 Chr.2: 113062460-113062460 on Build GRCh38
rs749973247 Chr.2: 113062438-113062438 on Build GRCh38
rs764733773 Chr.2: 113062271-113062271 on Build GRCh38
rs374135503 Chr.2: 113062387-113062387 on Build GRCh38
rs754925032 Chr.2: 113062285-113062285 on Build GRCh38
rs749941611 Chr.2: 113062443-113062446 on Build GRCh38
rs904500734 Chr.2: 113062477-113062477 on Build GRCh38
rs371695955 Chr.2: 113062423-113062423 on Build GRCh38
rs201346453 Chr.2: 113062470-113062470 on Build GRCh38
rs964943519 Chr.2: 113062376-113062376 on Build GRCh38
rs989206103 Chr.2: 113062407-113062407 on Build GRCh38
rs374849220 Chr.2: 113062447-113062447 on Build GRCh38
rs767374989 Chr.2: 113062453-113062453 on Build GRCh38
rs1047310672 Chr.2: 113062465-113062465 on Build GRCh38
rs754661425 Chr.2: 113062448-113062448 on Build GRCh38
rs780933637 Chr.2: 113062298-113062298 on Build GRCh38
rs749501046 Chr.2: 113062410-113062410 on Build GRCh38
rs367825442 Chr.2: 113062280-113062280 on Build GRCh38
rs910768228 Chr.2: 113062363-113062363 on Build GRCh38
rs553382111 Chr.2: 113062276-113062276 on Build GRCh38

View Full Product Details