Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 86152609-86152876 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6V0D2
Entrez Gene ID
245972
Gene Name
ATPase H+ transporting V0 subunit d2
Gene Aliases
ATP6D2, VMA6
UniGene
Hs.436360
Gene Chromosome Location
Chr.8: 86098910-86154225 on Build GRCh38
Transcripts
NM_152565.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6114212 c.898A>T p.M300L Chr.8: 86152822-86152822 on Build GRCh38
2789562 c.892-2A>G p.? Chr.8: 86152814-86152814 on Build GRCh38
2789563 c.897A>T p.Q299H Chr.8: 86152821-86152821 on Build GRCh38
454905 c.900G>C p.M300I Chr.8: 86152824-86152824 on Build GRCh38
5455691 c.934G>A p.G312S Chr.8: 86152858-86152858 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs540264817 Chr.8: 86152687-86152687 on Build GRCh38
rs901568753 Chr.8: 86152795-86152795 on Build GRCh38
rs758248296 Chr.8: 86152791-86152791 on Build GRCh38
rs747659877 Chr.8: 86152840-86152840 on Build GRCh38
rs10504820 Chr.8: 86152782-86152782 on Build GRCh38
rs765487536 Chr.8: 86152770-86152770 on Build GRCh38
rs888999517 Chr.8: 86152706-86152706 on Build GRCh38
rs998567000 Chr.8: 86152814-86152814 on Build GRCh38
rs528774808 Chr.8: 86152746-86152746 on Build GRCh38
rs901878560 Chr.8: 86152669-86152669 on Build GRCh38
rs779838695 Chr.8: 86152799-86152799 on Build GRCh38
rs746898746 Chr.8: 86152806-86152806 on Build GRCh38
rs970752253 Chr.8: 86152851-86152851 on Build GRCh38
rs754904689 Chr.8: 86152808-86152808 on Build GRCh38
rs142355302 Chr.8: 86152842-86152842 on Build GRCh38
rs780703101 Chr.8: 86152821-86152821 on Build GRCh38
rs961837886 Chr.8: 86152769-86152769 on Build GRCh38
rs552285877 Chr.8: 86152655-86152655 on Build GRCh38
rs1011685591 Chr.8: 86152831-86152831 on Build GRCh38
rs772641454 Chr.8: 86152835-86152841 on Build GRCh38
rs757035613 Chr.8: 86152845-86152845 on Build GRCh38
rs10095107 Chr.8: 86152657-86152657 on Build GRCh38
rs116131684 Chr.8: 86152726-86152726 on Build GRCh38
rs892818661 Chr.8: 86152824-86152824 on Build GRCh38
rs373357496 Chr.8: 86152697-86152697 on Build GRCh38
rs997923820 Chr.8: 86152671-86152671 on Build GRCh38

View Full Product Details