Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 3463299-3463524 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOK7
Entrez Gene ID
285489
Gene Name
docking protein 7
Gene Aliases
C4orf25, CMS10, CMS1B
UniGene
Hs.122110
Gene Chromosome Location
Chr.4: 3463306-3501476 on Build GRCh38
Transcripts
NM_001164673.1 NM_001301071.1 NM_173660.4 XM_011513435.2 XM_011513437.2 XM_017008038.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3603400 c.41G>A p.R14Q Chr.4: 3463416-3463416 on Build GRCh38
3603399 c.41G>A p.R14Q Chr.4: 3463416-3463416 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs906657133 Chr.4: 3463453-3463454 on Build GRCh38
rs768096493 Chr.4: 3463410-3463410 on Build GRCh38
rs762404326 Chr.4: 3463460-3463460 on Build GRCh38
rs863223277 Chr.4: 3463505-3463505 on Build GRCh38
rs757053604 Chr.4: 3463448-3463448 on Build GRCh38
rs976572936 Chr.4: 3463490-3463490 on Build GRCh38
rs746832145 Chr.4: 3463349-3463349 on Build GRCh38
rs745791856 Chr.4: 3463453-3463453 on Build GRCh38
rs375168516 Chr.4: 3463407-3463407 on Build GRCh38
rs769791490 Chr.4: 3463454-3463454 on Build GRCh38
rs572342022 Chr.4: 3463345-3463345 on Build GRCh38
rs762348331 Chr.4: 3463331-3463331 on Build GRCh38
rs767285554 Chr.4: 3463454-3463455 on Build GRCh38
rs753359348 Chr.4: 3463460-3463465 on Build GRCh38
rs886043943 Chr.4: 3463496-3463496 on Build GRCh38
rs751333072 Chr.4: 3463440-3463440 on Build GRCh38
rs763739780 Chr.4: 3463461-3463461 on Build GRCh38
rs756061120 Chr.4: 3463455-3463456 on Build GRCh38
rs768186113 Chr.4: 3463336-3463336 on Build GRCh38
rs752403967 Chr.4: 3463342-3463342 on Build GRCh38
rs1048076499 Chr.4: 3463360-3463360 on Build GRCh38
rs886038744 Chr.4: 3463493-3463493 on Build GRCh38
rs752213297 Chr.4: 3463427-3463427 on Build GRCh38
rs71180187 Chr.4: 3463461-3463462 on Build GRCh38
rs759348603 Chr.4: 3463452-3463453 on Build GRCh38
rs763233743 Chr.4: 3463382-3463382 on Build GRCh38
rs146168804 Chr.4: 3463346-3463346 on Build GRCh38
rs754565565 Chr.4: 3463341-3463341 on Build GRCh38
rs766545809 Chr.4: 3463451-3463453 on Build GRCh38
rs768798756 Chr.4: 3463402-3463402 on Build GRCh38
rs753349076 Chr.4: 3463340-3463340 on Build GRCh38
rs191800156 Chr.4: 3463370-3463370 on Build GRCh38
rs562528761 Chr.4: 3463459-3463459 on Build GRCh38
rs772575255 Chr.4: 3463457-3463457 on Build GRCh38
rs912327595 Chr.4: 3463442-3463442 on Build GRCh38
rs781051112 Chr.4: 3463452-3463452 on Build GRCh38
rs751188494 Chr.4: 3463463-3463463 on Build GRCh38
rs370421989 Chr.4: 3463406-3463406 on Build GRCh38
rs770809231 Chr.4: 3463352-3463352 on Build GRCh38
rs759054772 Chr.4: 3463420-3463420 on Build GRCh38
rs970805305 Chr.4: 3463339-3463339 on Build GRCh38
rs865893625 Chr.4: 3463450-3463450 on Build GRCh38
rs865811359 Chr.4: 3463412-3463412 on Build GRCh38
rs577687998 Chr.4: 3463437-3463437 on Build GRCh38
rs775604699 Chr.4: 3463371-3463371 on Build GRCh38
rs767030857 Chr.4: 3463458-3463458 on Build GRCh38
rs943989420 Chr.4: 3463462-3463462 on Build GRCh38
rs751937397 Chr.4: 3463328-3463328 on Build GRCh38
rs774468059 Chr.4: 3463456-3463456 on Build GRCh38
rs758049699 Chr.4: 3463429-3463429 on Build GRCh38
rs777859905 Chr.4: 3463457-3463458 on Build GRCh38
rs769850502 Chr.4: 3463443-3463457 on Build GRCh38
rs751749004 Chr.4: 3463451-3463456 on Build GRCh38
rs543525751 Chr.4: 3463455-3463455 on Build GRCh38
rs764824461 Chr.4: 3463421-3463421 on Build GRCh38
rs779230474 Chr.4: 3463451-3463452 on Build GRCh38
rs745885610 Chr.4: 3463366-3463366 on Build GRCh38
rs763022747 Chr.4: 3463451-3463451 on Build GRCh38
rs35131207 Chr.4: 3463470-3463470 on Build GRCh38
rs943831176 Chr.4: 3463435-3463435 on Build GRCh38
rs368106071 Chr.4: 3463411-3463411 on Build GRCh38
rs781143252 Chr.4: 3463357-3463357 on Build GRCh38
rs764508173 Chr.4: 3463326-3463326 on Build GRCh38

View Full Product Details