Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154442198-154442435 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDI1
Entrez Gene ID
2664
Gene Name
GDP dissociation inhibitor 1
Gene Aliases
1A, GDIL, MRX41, MRX48, OPHN2, RABGD1A, RABGDIA, XAP-4
UniGene
Hs.595950
Gene Chromosome Location
Chr.X: 154436913-154443467 on Build GRCh38
Transcripts
NM_001493.2

SNP Detail

SNP ID Chromosome Location
rs371655047 Chr.X: 154442374-154442374 on Build GRCh38
rs782263053 Chr.X: 154442384-154442384 on Build GRCh38
rs782789881 Chr.X: 154442331-154442331 on Build GRCh38
rs782440937 Chr.X: 154442236-154442236 on Build GRCh38
rs782055995 Chr.X: 154442369-154442370 on Build GRCh38
rs782004436 Chr.X: 154442266-154442266 on Build GRCh38
rs782068810 Chr.X: 154442290-154442290 on Build GRCh38
rs368434464 Chr.X: 154442354-154442354 on Build GRCh38
rs782787069 Chr.X: 154442367-154442367 on Build GRCh38
rs781969275 Chr.X: 154442326-154442326 on Build GRCh38
rs374368885 Chr.X: 154442267-154442267 on Build GRCh38
rs987108160 Chr.X: 154442351-154442351 on Build GRCh38
rs782018234 Chr.X: 154442280-154442280 on Build GRCh38
rs782549320 Chr.X: 154442357-154442357 on Build GRCh38
rs975406035 Chr.X: 154442264-154442264 on Build GRCh38
rs782655354 Chr.X: 154442397-154442397 on Build GRCh38
rs934237552 Chr.X: 154442323-154442323 on Build GRCh38
rs377631325 Chr.X: 154442268-154442268 on Build GRCh38
rs782232197 Chr.X: 154442241-154442241 on Build GRCh38
rs782556991 Chr.X: 154442369-154442369 on Build GRCh38
rs201330829 Chr.X: 154442301-154442301 on Build GRCh38
rs782501562 Chr.X: 154442386-154442386 on Build GRCh38
rs782346555 Chr.X: 154442249-154442249 on Build GRCh38
rs140855833 Chr.X: 154442404-154442404 on Build GRCh38
rs782402492 Chr.X: 154442223-154442223 on Build GRCh38
rs782600356 Chr.X: 154442378-154442378 on Build GRCh38
rs370898685 Chr.X: 154442277-154442277 on Build GRCh38

View Full Product Details